Merck
  • Sub-2-μm seeded growth mesoporous thin shell particles for high-performance liquid chromatography: Synthesis, functionalisation and characterisation.
Sub-2-μm seeded growth mesoporous thin shell particles for high-performance liquid chromatography: Synthesis, functionalisation and characterisation.
Journal of chromatography. A (2015-06-02)
Victor K Langsi, Benjamin A Ashu-Arrah, Jeremy D Glennon
ABSTRACT
Nanometer control over the porous shell thickness of sub-2-μm-shell particles is investigated. Three seeded growth mesoporous thin shell particles for HPLC were prepared, with 0.05μm (or 50nm) porous shell layers: particle sizes 1.5μm (solid core diameters 1.4μm), 1.7μm (solid core diameter 1.6μm), 1.9μm (solid core diameter 1.8μm) and compared with a fourth 1.7μm particle (solid core diameter 1.4μm) surrounded by 0.15μm (or 150nm) porous shell thickness. The thin shell particles were functionalised using a mono-functional octadecyldimethylchlorosilane ligand (C20H43SiCl) under optimised reflux conditions and packed in-house in narrow bore columns (2.1 I.D.×50mm) denoted as TS1.5-50-C18, TS1.7-50-C18, and TS1.9-50-C18 respectively. The synthesised thin shell particles and bonded materials were comprehensively characterised using scanning electron microscopy (SEM), transmission electron microscopy (TEM), dynamic light scattering (DLS), zeta potential, BET analysis, elemental analysis (CHN), thermogravimetric analysis (TGA) and diffuse reflectance infrared Fourier transform (DRIFT) spectroscopy. Experimental data from inverse size exclusion chromatography (ISEC) was used to measure external, internal and total column porosities. Five probe analytes (uracil, naphthalene, acetophenone, benzene and toluene) were chosen for the chromatographic performance analysis of these columns. Column evaluation and measurements of height equivalent to a theoretical plate (HETP) data were performed on naphthalene using 55% acetonitile in water. The retention coefficients for the thin shell particles (TS1.9-50-C18, TS1.7-50-C18, TS1.5-50-C18) were in the range 1.26-1.35 and 5.6 for the core-shell particle (EiS1.7-150-C18). The minimum reduced plate heights range from 3.89 to 4.26 for the thin shell particles and 2.03 for the core-shell particle.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Chloro(dimethyl)octadecylsilane, 95%
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Rhein
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer