Merck
  • A convenient screening method to differentiate phenolic skin whitening tyrosinase inhibitors from leukoderma-inducing phenols.
A convenient screening method to differentiate phenolic skin whitening tyrosinase inhibitors from leukoderma-inducing phenols.
Journal of dermatological science (2015-08-01)
Shosuke Ito, Kazumasa Wakamatsu
ABSTRACT
Tyrosinase is able to oxidize a great number of phenols and catechols to form ortho-quinones. Ortho-quinones are highly reactive compounds that exert cytotoxicity through binding with thiol enzymes and the production of reactive oxygen species. Certain phenolic (and catecholic) compounds are known to induce contact/occupational leukoderma through activation to ortho-quinones. We report a convenient screening method to follow the oxidation of those leukoderma-inducing phenols by mushroom tyrosinase. Oxidation of phenolic compounds by mushroom tyrosinase was followed periodically by UV-vis spectrophotometry. The production of ortho-quinones were confirmed by their absorptions around 400-420 nm. HPLC analysis after reduction with NaBH4 detected the corresponding catechols. Leukoderma-inducing phenols, rhododendrol, raspberry ketone, 4-methoxyphenol, 4-benzyloxyphenol, 4-tert-butylphenol, and 4-tert-butylcatechol, were readily oxidized by mushroom tyrosinase to form ortho-quinones. On the other hand, phenolic skin whitening tyrosinase inhibitors, ellagic acid, 4-n-butylresorcinol, potassium 4-methoxysalicylate, and 2,2'-dihydroxy-5,5'-di-n-propylbiphenyl, were not oxidized by mushroom tyrosinase, while arbutin was only slowly oxidized. This study has provided a convenient screening method to differentiate phenolic skin whitening tyrosinase inhibitors from leukoderma-inducing phenols. A common chemical feature of the latter group of compounds is that they are readily oxidized by tyrosinase to form reactive ortho-quinone species. The present results point out the necessity that tyrosinase inhibitors should also be examined as substrates if they are phenolic compounds.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
4-tert-Butylphenol, 99%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
Sigma-Aldrich
4-Propylphenol, 99%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%