Skip to Content
Merck
All Photos(1)

Key Documents

173401

Sigma-Aldrich

1,2,4-Benzenetriol

ReagentPlus®, 99%

Synonym(s):

Hydroxyhydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

mp

140 °C (subl.) (lit.)

SMILES string

Oc1ccc(O)c(O)c1

InChI

1S/C6H6O3/c7-4-1-2-5(8)6(9)3-4/h1-3,7-9H

InChI key

GGNQRNBDZQJCCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,4-Benzenetriol is also known as hydroxyhydroquinone. It is an intermediary metabolite of benzene that is present in roasted coffee beans. It is mutagenic and it causes cleaving of DNA single strands by the generation of reactive oxygen species.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of hydrogen peroxide from hydroxyhydroquinone and its inhibition by superoxide dismutase
Hiramoto K, et al.
Journal of Oleo Science, 50(1), 21-28 (2001)
DNA breakage induced by 1, 2, 4-benzenetriol: relative contributions of oxygen-derived active species and transition metal ions
Li AS, et al.
Free Radical Biology & Medicine, 30(9), 943-956 (2001)
Atsushi Suzuki et al.
American journal of hypertension, 20(5), 508-513 (2007-05-09)
Ferulic acid (FA), a phytochemical constituent, has antihypertensive effects, but a detailed understanding of its effects on vascular function remains unclear. The vasoreactivity of FA was assessed using aortic rings isolated from normotensive Wistar Kyoto (WKY) and spontaneously hypertensive rats
Zbigniew Srokol et al.
Carbohydrate research, 339(10), 1717-1726 (2004-06-29)
During the hydrothermal upgrading of biomass, hydrolysis to glucose is an important step. To elucidate some of the reaction pathways that follow this initial hydrolysis, the hydrothermal treatment (340 degrees C, 27.5 MPa, 25-204 s) of dilute (50 mM) solutions
Ryuji Ochiai et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 32(11), 969-974 (2009-08-29)
Recent studies suggest that chlorogenic acids, which are the main components of the polyphenol class in coffee, decrease blood pressure, and that hydroxyhydroquinone (HHQ), which is generated by roasting coffee beans, inhibits the antihypertensive effect of chlorogenic acids in brewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service