Merck
  • Soluble Phenolic Compounds in Different Cultivars of Red Clover and Alfalfa, and their Implication for Protection against Proteolysis and Ammonia Production in Ruminants.
Soluble Phenolic Compounds in Different Cultivars of Red Clover and Alfalfa, and their Implication for Protection against Proteolysis and Ammonia Production in Ruminants.
Natural product communications (2015-09-29)
Isabelle A Kagan, Ben M Goff, Michael D Flythe
ABSTRACT
Red clover (Trifolium pratense) contains soluble phenolic compounds with roles in inhibiting proteolysis and ammonia production. Alfalfa (Medicago sativa) has been found to have a low phenolic content, but few alfalfa and red clover cultivars have been compared for phenolic content. Total soluble phenolics were quantified by a Folin-Ciocalteu colorimetric assay in nine red clover and 27 alfalfa cultivars. Mean total phenolic contents of red clover and alfalfa were 36.5 ± 4.3 mg/gdw and 15.8 ± 1.4 mg/gdw, respectively, with the greater standard deviation of red clover possibly indicating more diversity in phenolic content. Because different phenolic standards had different response factors in the colorimetric assay, the red clover and 11 alfalfa cultivars were analyzed by HPLC to determine if the differences in total soluble phenolics between genera reflected differences in the amounts of phenolics or in the classes of phenolics responding to the colorimetric assay. Two red clover cultivars differed in total phenolics and phaselic acid. Alfalfa produced different phenolic compounds from red clover, at lower concentrations. Extracts of two red clover cultivars were separated by thin-layer chromatography (TLC), and the bands were assayed for activity against Clostridium sticklandii, a bovine ruminal hyper ammonia-producing bacterium (HAB). Only biochanin A had anti-HAB activity. Inhibitory amounts indicated that five red clover cultivars could be suitable sources of anti-HAB activity.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Ononin, ≥99.0% (TLC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Formononetin, ≥99.0% (TLC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate