Merck
  • Quantification of remifentanil and propofol in human plasma: a LC-MS/MS assay validated according to the EMA guideline.
Quantification of remifentanil and propofol in human plasma: a LC-MS/MS assay validated according to the EMA guideline.
Bioanalysis (2015-08-01)
Jean Claude Alvarez, Emuri Abe, Isabelle Etting, Morgan Le Guen, Philippe Devillier, Stanislas Grassin-Delyle
ABSTRACT
Remifentanil and propofol are often used in combination for general anesthesia. We developed a method using LC-MS for their simultaneous measurement in human plasma. Methodology & results: After addition of remifentanil-(13)C6 and propofol-d18 (IS), 500 µl of plasma were extracted with ethylacetate/hexane. Analysis conditions included gradient elution (water/acetonitrile), electrospray ionization and detection with a triple quadripole mass spectrometer. Remifentanil and propofol were ionized in the positive and negative mode, respectively. The method was validated according to the European Medicines Agency guideline for the validation of bioanalytical methods, then successfully applied to clinical samples from three patients who had undergone liver transplantation. This method is suitable for the simultaneous quantification of remifentanil and propofol in clinical studies.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
2-Ethylaniline, 98%
Sigma-Aldrich
Sodium carbonate decahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, 99%