Merck
  • Whole Grain Consumption Increases Gastrointestinal Content of Sulfate-Conjugated Oxylipins in Pigs - A Multicompartmental Metabolomics Study.
Whole Grain Consumption Increases Gastrointestinal Content of Sulfate-Conjugated Oxylipins in Pigs - A Multicompartmental Metabolomics Study.
Journal of proteome research (2015-07-03)
Anne Krog Ingerslev, Ibrahim Karaman, Murat Bağcıoğlu, Achim Kohler, Peter Kappel Theil, Knud Erik Bach Knudsen, Mette Skou Hedemann
ABSTRACT
The effects of increased intake of dietary fiber as either whole grain rye or as resistant starch compared to a typical low dietary fiber Western-style diet on the metabolomics responses were studied in gastrointestinal content and tissue, peripheral plasma, and urine using a multicompartmental nontargeted liquid chromatography-mass spectrometry (LC-MS) approach in pigs. Both unsupervised and supervised multivariate analyses were used to study differences between the intervention groups, revealing significant effects of the dietary intervention on gastrointestinal contents and urine metabolites. Consumption of a diet composed of whole grain rye flakes and enzyme treated wheat bran increased gastrointestinal content of novel sulfate- and acetic acid-conjugated oxylipins, and urinary secretion of phenolic compounds. Furthermore, potential gastrointestinal and urinary biomarkers for consumption of resistant starch, whole grain rye, and a low dietary fiber intake were detected using multiblock analysis. This study provides insight into microbial fermentation products in the gastrointestinal tract and suggests a potential role in sulfate conjugation of metabolites on the bioavailability of ingested nutrients.
Sigma-Aldrich
Glycine-1-13C, 99 atom % 13C
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%
Sigma-Aldrich
Hyodeoxycholic acid, ≥98%
Sigma-Aldrich
Glycine-1-13C, endotoxin tested, 99 atom % 13C
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Suberic acid, 98%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Azelaic acid, technical grade, 80%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Enterolactone, ~95% (HPLC)
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Azelaic acid, 98%
Sigma-Aldrich
Hexachloro-2-propanone, 99%
Sigma-Aldrich
4-Pyridoxic acid, ≥98%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sebacic acid, ≥95.0% (GC)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexachloro-2-propanone, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Sebacic acid, 99%
Sigma-Aldrich
Citric acid, 99%