Skip to Content
Merck
All Photos(1)

Key Documents

467804

Sigma-Aldrich

Hydroxylamine solution

50 wt. % in H2O, 99.999%

Synonym(s):

HDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH
CAS Number:
Molecular Weight:
33.03
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99.999%

concentration

50 wt. % in H2O

bp

107 °C

density

1.078 g/mL at 25 °C

SMILES string

NO

InChI

1S/H3NO/c1-2/h2H,1H2

InChI key

AVXURJPOCDRRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxylamine solution is widely used as a reducing agent and antioxidant in organic synthesis, polymer industry and water treatment.

Application

Hydroxylamine solution (NH2OH) can be used as a reactant for the preparation of:
  • Primary amides from aldehydes catalyzed by an arene–ruthenium(II) complex.
  • Hydroxyaminoguanidines and carboxamide derivatives of ofloxacin for biological studies.
  • Fe3O4/Au (GoldMag) nanoparticles for antibody immobilization.

Reactant for preparation of:
  • Prodrug for cardiovascular agent Nω-hydroxy-L-arginine (NOHA, nitric oxide precursor)
  • Hydroxyaminoguanidines as anti-cancer agents
  • Nonsteroidal 2,3-dihydroquinoline glucocorticoid receptor agonists with reduced phosphoenolpyruvate caboxykinase (PEPCK) transactivation
  • Carboxamide derivatives of ofloxacin with improved antimicrobial properties
  • Analogues of coumarin based TNF-α converting enzyme (TACE) inhibitors
  • HIV integrase inhibitors

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Desen. Expl. 4 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Blood, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Hydroxy-N′-aminoguanidines as anti-cancer lead molecule: QSAR, synthesis and biological evaluation
Basu A, et al.
Bioorganic & Medicinal Chemistry Letters, 21(11), 3324-3328 (2011)
Ruthenium-catalyzed one-pot synthesis of primary amides from aldehydes in water
Garcia-Alvarez R, et al.
Royal Society of Chemistry Advances, 3(17), 5889-5894 (2013)
The synthesis of GoldMag nano-particles and their application for antibody immobilization
Cui Y, et al.
Biomedical Microdevices, 7(2), 153-156 (2005)
Rika Goda et al.
Drug metabolism and disposition: the biological fate of chemicals, 34(5), 828-835 (2006-03-02)
Flutamide (2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]-propanamide), a nonsteroidal antiandrogen, is used in the treatment of prostate cancer but is occasionally associated with hepatic dysfunction. In the present study, the metabolism of flutamide including the formation of the possible reactive toxic metabolites was investigated using
Synthesis, characterization and biological activity of a series of carboxamide derivatives of ofloxacin
Arayne MS, et al.
Archives of Pharmacal Research, 33(12), 1901-1909 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service