Merck
  • Activation of Flucloxacillin-Specific CD8+ T-Cells With the Potential to Promote Hepatocyte Cytotoxicity in a Mouse Model.
Activation of Flucloxacillin-Specific CD8+ T-Cells With the Potential to Promote Hepatocyte Cytotoxicity in a Mouse Model.
Toxicological sciences : an official journal of the Society of Toxicology (2015-04-17)
Ryan Nattrass, Lee Faulkner, Marc Vocanson, Daniel J Antoine, Anja Kipar, Gerry Kenna, Jean-Francois Nicolas, B Kevin Park, Dean J Naisbitt
ABSTRACT
There are currently no animal models of drug-induced liver injury (DILI) where the adaptive immune system has been shown to damage the liver. Thus, it is difficult to explore the mechanistic basis of the tissue injury. The aim of this study was to use C57BL/6 CD4+-deficient mice with a mutation in the αβ gene encoding for Major histocompatibilty complex (MHC) class II molecules to (1) develop a mouse model of flucloxacillin sensitization, (2) explore whether drug-specific CD8+ kill primary hepatocytes, and (3) analyze perturbations in liver integrity following oral exposure to flucloxacillin. CD8+ T-cells from lymph nodes of flucloxacillin-sensitized mice were stimulated to proliferate, secrete interferon (IFN-γ) and granzyme B, and induce hepatocyte apoptosis in a concentration-dependent manner following ex vivo stimulation. The T-cell response was antigen-specific; T-cells were not activated with other β-lactam antibiotics. Furthermore, T-cell responses only occurred in the presence of flucloxacillin-pulsed antigen presenting cells. In separate experiments, flucloxacillin-specific T-cells were induced to migrate to the mesenteric lymph nodes using retinoic acid, prior to administration of oral flucloxacillin, and analysis of plasma biomarkers of liver injury. Oral exposure to flucloxacillin resulted in mild elevations in alanine aminotransferase, liver, and gall bladder leukocyte infiltration and a marked swelling of the gall bladder. Thus, CD4+-deficient mice represent a promising model to study the role of the adaptive immune system in DILI.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Flucloxacillin sodium, ≥95% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%