Merck
  • Labile ruthenium(ii) complexes with extended phenyl-substituted terpyridyl ligands: synthesis, aquation and anticancer evaluation.
Labile ruthenium(ii) complexes with extended phenyl-substituted terpyridyl ligands: synthesis, aquation and anticancer evaluation.
Dalton transactions (Cambridge, England : 2003) (2015-08-08)
Huaiyi Huang, Pingyu Zhang, Yu Chen, Liangnian Ji, Hui Chao
ABSTRACT
Ruthenium complexes have been considered as promising substitutes for cisplatin in cancer chemotherapy. However, novel ruthenium-based therapies are faced with some limitations, such as unimpressive cytotoxicity toward solid tumors. Herein, we designed and synthesized phenyl-substituted terpyridyl ruthenium(ii) complexes ([Ru(tpy)(bpy)Cl](+) (Ru1), [Ru(phtpy)(bpy)Cl](+) (Ru2) and [Ru(biphtpy)(bpy)Cl](+) (Ru3)) which exhibited distinctly different anticancer activity. Ru1-Ru3 all underwent moderate aquation in buffer solution and this process was significantly inhibited by high chloride concentration. Cancer cells were found to readily uptake the relatively hydrophobic Ru3, as quantified using inductively coupled plasma mass spectrometry (ICP-MS). Ru1 was found to be non-cytotoxic (IC50 > 100 μM) while Ru3 exhibited very promising cytotoxicity on both two-dimensional (2D) cancer cell monolayers and 3D MCTSs. An antiproliferative assay revealed that Ru3 significantly inhibited cellular DNA replication which ultimately induced apoptosis of cancer cells.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
Ruthenium on carbon, extent of labeling: 5 wt. % loading
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Ruthenium(III) chloride hydrate, 38.0-42.0% Ru basis
Sigma-Aldrich
Ruthenium(III) chloride hydrate, 99.98% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Ruthenium, Ruthenium, bar, 50mm x 2mm x 2mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.1μm, specific density 122μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, foil, 10x10mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, pellets, 2.5g, max. size 10mm, 99.9%
Ruthenium, Ruthenium, foil, 25x25mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, foil, 6x6mm, thickness 1.0mm, 99.9%
Ruthenium, Ruthenium, rod, 12.7mm, diameter 12.7mm, 99.9%
Ruthenium, Ruthenium, microfoil, disks, 10mm, thinness 0.025μm, specific density 30.5μg/cm2, permanent mylar 3.5μm support, 99.9%
Ruthenium, Ruthenium, pellets, 5g, max. size 10mm, 99.9%