Skip to Content
Merck
All Photos(3)

Key Documents

P1525

Sigma-Aldrich

trans-Platinum(II)diammine dichloride

Synonym(s):

trans-Diamminedichloroplatinum(II), trans-Dichlorodiammineplatinum(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

mp

340 °C (dec.) (lit.)

SMILES string

N.N.Cl[Pt]Cl

InChI

1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2

InChI key

LXZZYRPGZAFOLE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian G Hartinger et al.
Inorganic chemistry, 47(1), 17-19 (2007-12-11)
A proof-of-principle study on the application of a top-down electrospray ionization Fourier transform ion cyclotron resonance mass spectrometric approach for characterization of the primary binding sites of the platinum anticancer agents cisplatin, transplatin, and oxaliplatin on ubiquitin is presented. Through
Leticia Cubo et al.
Dalton transactions (Cambridge, England : 2003), (18)(18), 3457-3466 (2009-04-22)
Three (15)N-labelled trans-Pt(ii) amine complexes with isopropylamine ((15)N-ipa), methylamine ((15)N-ma) and dimethylamine ((15)N-dma) have been prepared and characterized. 2D [(1)H,(15)N] HSQC NMR spectroscopy was used to obtain the rate and equilibrium constants for the aquation of trans-[PtCl(2)((15)N-ipa)((15)N-ma)] ((15)N-1), trans-[PtCl(2)((15)N-dma)((15)N-ma)] ((15)N-2)
Andrei V Karotki et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 14(7), 1129-1138 (2009-06-19)
Human metallothioneins, small cysteine- and metal-rich proteins, play an important role in the acquired resistance to platinum-based anticancer drugs. These proteins contain a M(II)4(CysS)11 cluster and a M(II)3(CysS)9 cluster localized in the alpha-domain and the beta-domain, respectively. The noninducible isoform
Leticia Cubo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(36), 9139-9146 (2009-08-06)
A novel trans-platinum(II) complex bearing one dimethylamine (dma) and one methylamine (ma) ligand, namely trans-[PtCl(2)(dma)(ma)], recently synthesised and characterised in our laboratory, displayed relevant antiproliferative properties in vitro, being more active than the parent complex, trans-[PtCl(2)(dma)(ipa)], which has isopropylamine (ipa)
Angela Casini et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 14(5), 761-770 (2009-03-17)
Electrospray ionisation mass spectrometry was used to analyse the reactions of metal compounds with mixtures of selected proteins. Three representative medicinally relevant compounds, cisplatin, transplatin and the organometallic ruthenium compound RAPTA-C, were reacted with a pool of three proteins, ubiquitin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service