Merck
  • Adenovirus-mediated p53 and ING4 gene co-transfer elicits synergistic antitumor effects through enhancement of p53 acetylation in breast cancer.
Adenovirus-mediated p53 and ING4 gene co-transfer elicits synergistic antitumor effects through enhancement of p53 acetylation in breast cancer.
Oncology reports (2015-11-05)
Jie Wu, Yanbo Zhu, Chun Xu, Hong Xu, Xiumin Zhou, Jicheng Yang, Yufeng Xie, Min Tao
ABSTRACT
Multigene-based combination therapy may be an effective practice in cancer gene therapy. Substantial studies have demonstrated that tumor suppressor p53 acetylation is indispensable for p53 activation. Inhibitor of growth 4 (ING4), as a novel tumor suppressor, is capable of remarkably enhancing p53 acetylation and its transcriptional activity. Hence, we assumed that combined treatment of p53 and ING4 double tumor suppressors would exhibit enhanced antitumor effects. The combined therapeutic efficacy of p53 and ING4 for human cancers has not been previously reported. We thus generated multiple promoter expression cassette-based recombinant adenovirus-co-expressing ING4 and p53 double tumor suppressor genes (AdVING4/p53), evaluated the combined effects of AdVING4/p53 on breast cancer using the MDA-MB-231 (mutant p53) human breast cancer cell line, and also elucidated its underlying molecular mechanisms. We demonstrated that AdVING4/p53-mediated p53 and ING4 co-expression induced synergistic growth inhibition and apoptosis as well as enhanced effects on upregulation of acetylated p53, P21, Bax, PUMA, Noxa, cleaved caspase-9, cleaved caspase-3 and cleaved PARP, and downregulation of Bcl-2, CD31 and microvessel density (MVD) in MDA-MB-231 breast cancer in vitro and/or in vivo subcutaneous (s.c.) xenografted tumors. The synergistic antitumor activity elicited by AdVING4/p53 was closely associated with the enhanced activation of the intrinsic apoptotic pathway and synergistic inhibition of tumor angiogenesis, very possibly via ING4-mediated enhancement of p53 acetylation and activity. Thus, our results indicate that cancer gene therapy combining two or more tumor suppressors such as p53 and ING4 may constitute a novel and effective therapeutic modality for human breast cancer and other cancers.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis