Skip to Content
Merck
All Photos(5)

Key Documents

C3139

Sigma-Aldrich

Cesium chloride

optical grade, ≥99.5% trace metals basis

Synonym(s):

Caesium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

optical grade

Quality Level

Assay

≥99.5% trace metals basis

form

powder

mp

645 °C (lit.)

absorption

≤0.02 at 280 in H2O at 50%

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cesium chloride can be used:
  • To prepare Ga2S3-GeS2-CsCl glasses, which are transparent in the entire visible range.
  • To fabricate WO3 islands and network structures, which improves the light extraction efficiency of an OLED.
It can also be used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Quality

Yields a clear colorless solution.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Totally visible transparent chloro?sulphide glasses based on Ga2S3?GeS2-CsCl
Ledemi Y., et al.
Journal of Optoelectronics and Advanced Materials, 9, 3751-3751 (2007)
S J Garger et al.
Biochemical and biophysical research communications, 117(3), 835-842 (1983-12-28)
A procedure for rapid, preparative purification of plasmid DNA is described and compared with a conventional equilibrium centrifugation method. A discontinuous, two-step CsCl-ethidium bromide gradient is used, with the starting position of the plasmid-containing extract being at the bottom of
Enhancing light extraction efficiency of organic light-emitting diodes by embedding tungsten trioxide islands or network structure pattern-transferred from a self-assembled deliquesce cesium chloride mask
Bao Dong To, et al.
Organic Electronics, 53, 160-164 (2008)
Tuan A Ho et al.
Langmuir : the ACS journal of surfaces and colloids, 28(2), 1256-1266 (2011-12-14)
All-atom molecular dynamics simulations were conducted to study the dynamics of aqueous electrolyte solutions confined in slit-shaped silica nanopores of various degrees of protonation. Five degrees of protonation were prepared by randomly removing surface hydrogen atoms from fully protonated crystalline
Travis A Hage et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(8), 2714-2721 (2012-02-24)
We report a novel coupled system of sodium-activated potassium currents (I(KNa)) and persistent sodium currents (I(NaP)), the components of which are widely distributed throughout the brain. Its existence and importance has not been previously recognized. Although I(KNa) was known to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service