Merck
  • Mechanism of water oxidation by non-heme iron catalysts when driven with sodium periodate.
Mechanism of water oxidation by non-heme iron catalysts when driven with sodium periodate.
Dalton transactions (Cambridge, England : 2003) (2014-07-09)
Alexander R Parent, Takashi Nakazono, Shu Lin, Satoshi Utsunomiya, Ken Sakai
ABSTRACT
Iron tris(2-methylpyridyl)amine (TPA) and iron 1-(bis(2-methylpyridyl)amino)-2-methyl-2-propanoate (BPyA) salts are characterized as water oxidation catalysts (WOCs) using sodium periodate. Under the conditions used, these complexes serve as homogeneous WOCs as demonstrated via kinetic analysis and dynamic light scattering (DLS). The Fe(BPyA) salt serves as both a mononuclear and dinuclear catalyst, with the mononuclear form showing higher catalytic activity. Based on the H/D kinetic isotope effect and pH dependence, the rate determining step (RDS) in water oxidation (WO) by Fe(BPyA) is nucleophilic attack by water during O-O bond formation. In contrast, Fe(TPA) shows complex kinetic behavior due to the formation of multiple oxidation states of the complex in solution, each of which exhibits catalytic activity for WO. The RDS in WO by Fe(TPA) follows an equilibrium established between monomeric and dimeric forms of the catalyst. Under acidic conditions formation of the monomer is favored, which leads to an increase in the WO rate.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
2-Pyridinecarboxaldehyde, 99%
Sigma-Aldrich
2-(Chloromethyl)pyridine hydrochloride, 98%
Sigma-Aldrich
1-Amino-2-methyl-2-propanol, 95% anhydrous basis