Skip to Content
Merck
All Photos(1)

Key Documents

296945

Sigma-Aldrich

Sodium cyanoborohydride solution

5.0 M in 1 M NaOH

Synonym(s):

Sodium borocyanohydride, Sodium cyanoborohydride, Sodium cyanotrihydroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH3CN
CAS Number:
Molecular Weight:
62.84
MDL number:
UNSPSC Code:
12352306
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reagent type: reductant

concentration

5.0 M in 1 M NaOH

density

1.083 g/mL at 25 °C

SMILES string

[Na+].[H][B-]([H])([H])C#N

InChI

1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1

InChI key

CVDUGUOQTVTBJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium cyanoborohydride is a selective, mild reducing reagent for a number of functional groups such as aldehydes, ketones, acetals, epoxides and imines.

Application

Reactant for:
  • Direct conversion of heterocyclic aldehydes to esters (oxidizing behavior)
  • Reductive amination reactions
  • Intramolecular reductive cyclization reactions
  • Reductions reactions
  • Preparation of dicyanoborate-based ionic liquids as hypergolic fluids for bipropellants

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

e-EROS Encyclopedia of Reagents for Organic Synthesis (2016)
Franklin Afinjuomo et al.
Pharmaceutics, 11(5) (2019-05-28)
The propensity of monocytes to migrate into sites of mycobacterium tuberculosis (TB) infection and then become infected themselves makes them potential targets for delivery of drugs intracellularly to the tubercle bacilli reservoir. Conventional TB drugs are less effective because of
Shinsuke Sando et al.
Bioorganic & medicinal chemistry letters, 13(16), 2633-2636 (2003-07-23)
We report here that unprotected carbohydrates (maltose, lactose, cellobiose, and maltoheptaose) can be attached to the aminoalkylated oligonucleotides under mild reductive-amination conditions (aqueous borate buffer, pH 8.0, NaBH(3)CN, 60 degrees C) without notable side reactions. Quadruplex-forming G-rich oligonucleotide, 5'-aminoalkyl d(TGGGGT)
Jue-Liang Hsu et al.
Electrophoresis, 27(18), 3652-3660 (2006-08-24)
Stable-isotope labeling coupled with liquid-phase separation and MS analysis is a powerful technique for comparative proteomics. We developed a dimethyl labeling strategy (Anal. Chem. 2003, 75, 6843-6852 and J. Proteome Res. 2005, 4, 101-108) to label peptide N-terminus and epsilon-amino
Paul J Boersema et al.
Proteomics, 8(22), 4624-4632 (2008-10-14)
Stable isotope labeling is at present one of the most powerful methods in quantitative proteomics. Stable isotope labeling has been performed at both the protein as well as the peptide level using either metabolic or chemical labeling. Here, we present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service