Skip to Content
Merck
All Photos(1)

Key Documents

516813

Sigma-Aldrich

Hydrogen peroxide solution

50 wt. % in H2O, stabilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
EC Number:
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21

Agency

suitable for EPA 200.7

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

Assay

48-52% H2O2 basis (by KMnO4, titration)

form

liquid

reaction suitability

reaction type: C-H Activation
reagent type: catalyst
reagent type: oxidant

concentration

50 wt. % in H2O

mp

−40 °C

density

1.197 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a 50% solution of hydrogen peroxide in water. Hydrogen peroxide is more commonly being used as a green oxidizing agent. It is a better alternative to other oxidants because it has high active oxygen content and the by-product is water.

Application

Hydrogen peroxide solution may be used as an oxidizing agent in the following reactions:
  • Epoxidation of alkenes in the presence of homogeneous metal catalyst.
  • Oxidative halogenation in the presence of metal catalyst.
  • Oxidation of sulfides to sulfoxides and sulfones in the presence of polymer immobilised catalyst.

Other Notes

Contains proprietary inorganic tin-based stabilizer

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Ox. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent developments on the epoxidation of alkenes using hydrogen peroxide as an oxidant.
Grigoropoulou G, et al.
Green Chemistry, 5(1), 1-7 (2003)
Efficient and selective hydrogen peroxide-mediated oxidation of sulfides in batch and segmented and continuous flow using a peroxometalate-based polymer immobilised ionic liquid phase catalyst.
Doherty S, et al.
Green Chemistry, 17(3), 1559-1571 (2015)
Oxidative halogenation with ?green? oxidants: oxygen and hydrogen peroxide.
Podgorsek A, et al.
Angewandte Chemie (International Edition in English), 48(45), 8424-8450 (2009)
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
James R Van Brocklyn et al.
Cancer research, 74(19), 5364-5370 (2014-08-12)
Glioblastoma remains a devastating disease for which novel therapies are urgently needed. Here, we report that the Aurora-A kinase inhibitor alisertib exhibits potent efficacy against glioblastoma neurosphere tumor stem-like cells in vitro and in vivo. Many glioblastoma neurosphere cells treated

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service