Merck
  • Preparation and chromatographic evaluation of a newly designed steviol glycoside modified-silica stationary phase in hydrophilic interaction liquid chromatography and reversed phase liquid chromatography.
Preparation and chromatographic evaluation of a newly designed steviol glycoside modified-silica stationary phase in hydrophilic interaction liquid chromatography and reversed phase liquid chromatography.
Journal of chromatography. A (2015-03-03)
Tu Liang, Qing Fu, Aijin Shen, Hui Wang, Yu Jin, Huaxia Xin, Yanxiong Ke, Zhimou Guo, Xinmiao Liang
ABSTRACT
A diterpene glycoside compound, rebaudioside A (commonly abbreviated as RA), was immobilized onto porous silica surface through "thiol-ene" click chemistry strategy. The successful immobilization of the RA on the silica support was confirmed by FT-IR and elemental analysis. Chromatographic characteristics of the new stationary phase, named Click TE-RA, were evaluated by a set of diverse analytes such as carbohydrates, nucleosides, and organic acids in hydrophilic interaction liquid chromatography (HILIC) mode. The effects of water content, buffer pH and concentration were investigated and a typical HILIC retention feature of Click TE-RA was observed at high organic modifier content. The Click TE-RA stationary phase was further studied by a series of glycoside compounds. Tunable retention mechanisms from hydrophilic to hydrophobic interactions were observed. Separation of very polar compounds including oligosaccharides, nucleic acid bases and nucleosides using Click TE-RA in HILIC mode was successfully accomplished. In addition, separation of saponins both in HILIC and reversed-phase liquid chromatography (RPLC) modes was performed, demonstrating the presence of orthogonality between two different modes on Click TE-RA column. The multiple interactions induced by polar sugar group and hydrophobic aglycone group allowed this Click TE-RA to serve as a multi-mode stationary phase in two-dimensional liquid chromatography.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Cytosine, ≥99%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), purum, ≥98.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Cytidine, 99%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Orotic acid, ≥98% (titration), anhydrous
Sigma-Aldrich
Thymine, suitable for cell culture, BioReagent
Sigma-Aldrich
Uridine, ≥99%
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Thymine(Zidovudine Related Compound C), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Xylitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
Xylitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material