Skip to Content
Merck
All Photos(3)

Key Documents

179337

Sigma-Aldrich

Methanol

≥99.8%, ACS reagent, suitable for EPA 1613

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39020101
PubChem Substance ID:
NACRES:
NA.21

product name

Methanol, ACS reagent, ≥99.8%

grade

ACS reagent

Quality Level

100
200

Agency

suitable for EPA 1613

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

impurities

H2SO4, passes test (darkened)
MnO4- reducers, passes test
≤0.0002 meq/g Titr. base
≤0.0003 meq/g Titr. acid
≤0.001% acetaldehyde
≤0.001% acetone
≤0.001% formaldehyde
≤0.1% water

evapn. residue

≤0.001%

color

APHA: ≤10
clear

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol is an organic solvent that can be synthesized from syngas in the presence of CuO/ZnO/Al2O3 catalysts. It is an ideal candidate as a hydrogen source in fuel cell technology due to its high H/C ratio, low propensity for soot generation, relatively low reforming temperature and as it exists in liquid state at room temperature. In a direct methanol fuel cell (DMFC), methanol undergoes oxidation with air to generate electricity. The olefins (ethylene or propylene) formed from methanol via MTO (methanol-to-olefins) process, can be an alternative to oil and gas to produce hydrocarbon fuels.
Methanol undergoes thermochemical conversion to C2-C10 hydrocarbons in the presence of shape-selective zeolites has been reported. Its oxidation on Ru-Pt catalyst system by ruthenium ad-atoms has been proposed.

Application

Methanol has been used as a solvent for the sodium dodecyl sulfate-polyacrylamide gel electrophoretic (SDS-PAGE) analysis of alkaline protease isoforms isolated from larva of the spotted sand bass (Paralabrax maculatofasciatus). It has been used in the isolation and quantitative determination of carotenes and xanthophylls from Capsicum annum pericarp extracts by HPLC.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beyond oil and gas: the methanol economy.
Olah GA.
Angewandte Chemie (International Edition in English), 44(18) (2005)
Development of digestive enzyme activity in larvae of spotted sand bass Paralabrax maculatofasciatus II: electrophoretic analysis.
Alvarez-Gonzalez CA, et al.
Fish Physiology and Biochemistry, 36(1), 29-37 (2010)
Catalytic carbon dioxide hydrogenation to methanol: A review of recent studies.
Jadhav SG, et al.
Chemical Engineering Research and Design, 92(11), 2557-2567 (2014)
Methanol steam reforming for hydrogen generation via conventional and membrane reactors: a review.
Iulianelli A, et al.
Renewable and Sustainable Energy Reviews, 29, 355-368 (2014)
Review of methanol reforming-Cu-based catalysts, surface reaction mechanisms, and reaction schemes.
Yong ST, et al.
International Journal of Hydrogen Energy, 38(22), 9541-9552 (2013)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service