Merck
  • Novel recombinant multiepitope proteins for the diagnosis of asymptomatic leishmania infantum-infected dogs.
Novel recombinant multiepitope proteins for the diagnosis of asymptomatic leishmania infantum-infected dogs.
PLoS neglected tropical diseases (2015-01-09)
Angélica Rosa Faria, Luciano de Castro Veloso, Wendel Coura-Vital, Alexandre Barbosa Reis, Leonardo Miranda Damasceno, Ricardo T Gazzinelli, Hélida M Andrade
ABSTRACT
Visceral leishmaniasis is the most severe form of leishmaniasis. Worldwide, approximately 20% of zoonotic human visceral leishmaniasis is caused by Leishmania infantum, also known as Leishmania chagasi in Latin America. Current diagnostic methods are not accurate enough to identify Leishmania-infected animals and may compromise the effectiveness of disease control. Therefore, we aimed to produce and test two recombinant multiepitope proteins as a means to improve and increase accuracy in the diagnosis of canine visceral leishmaniasis (CVL). Ten antigenic peptides were identified by CVL ELISA in previous work. In the current proposal, the coding sequences of these ten peptides were assembled into a synthetic gene. Furthermore, other twenty peptides were selected from work by our group where good B and T cell epitopes were mapped. The coding sequences of these peptides were also assembled into a synthetic gene. Both genes have been cloned and expressed in Escherichia coli, producing two multiepitope recombinant proteins, PQ10 and PQ20. These antigens have been used in CVL ELISA and were able to identify asymptomatic dogs (80%) more effectively than EIE-LVC kit, produced by Bio-Manguinhos (0%) and DPP kit (10%). Moreover, our recombinant proteins presented an early detection (before PCR) of infected dogs, with positivities ranging from 23% to 65%, depending on the phase of infection in which sera were acquired. Our study shows that ELISA using the multiepitope proteins PQ10 and PQ20 has great potential in early CVL diagnosis. The use of these proteins in other methodologies, such as immunochromatographic tests, could be beneficial mainly for the detection of asymptomatic dogs.
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside