Skip to Content
Merck
All Photos(4)

Key Documents

60220

Sigma-Aldrich

Potassium phosphate monobasic

buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous
puriss. p.a.

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

Assay

99.5-100.5%

form

crystalline

impurities

≤0.001% total nitrogen (N)

loss

≤0.2% loss on drying, 105 °C

pH

4.2-4.5 (20 °C, 5%)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

solubility

water: 0.5 g/10 mL, clear, colorless to light yellow

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤30 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤15 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤0.001% (by ICP)

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate is widely used in buffers to prepare biological samples and examine biological interactions.

Application

Potassium phosphate monobasic has been used:
  • to prepare the working luminol solution to study the prooxidant activity in the chemiluminescent system luminol–H2O2
  • in phosphate buffer saline (PBS) to culture embryonic stem cells (ESCs) and generate embryoid bodies
  • in PBS to prepare polydimethylsiloxane (PDMS) substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prooxidant potential of \_2 nanoparticles towards hydrogen peroxide
Sozarukova MM, et al.
Nanosystems: Physics, Chemistry, Mathematics, 12(3), 283-290 (2021)
Jules D P Valentin et al.
Journal of colloid and interface science, 552, 247-257 (2019-05-28)
Many materials used in the medical settings such as catheters and contact lenses as well as most biological tissues are not purely elastic, but rather viscoelastic. While substrate elasticity has been investigated for its influence on bacterial adhesion, the impact
John E Schiel et al.
Talanta, 65(2), 495-500 (2008-10-31)
Potassium phosphate buffer is often used in methods such as equilibrium dialysis, high performance liquid chromatography (HPLC), and affinity capillary electrophoresis (ACE) for characterizing the binding of drugs and hormones with proteins or other ligands within the body. In these
Sonal Saxena et al.
MethodsX, 7, 101073-101073 (2020-10-22)
Investigation on the effects of disease-associated mutations on neurodevelopment is an essential approach to understand the molecular basis of neurological disorders and can be achieved by generating suitable animal models. However, some of the mutations preclude development of animal models
Casper Skovgaard et al.
Journal of applied physiology (Bethesda, Md. : 1985), 117(10), 1097-1109 (2014-09-06)
The purpose of this study was to examine whether speed endurance training (SET, repeated 30-s sprints) and heavy resistance training (HRT, 80-90% of 1 repetition maximum) performed in succession are compatible and lead to performance improvements in moderately trained endurance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service