Skip to Content
Merck
All Photos(5)

Key Documents

P0662

Sigma-Aldrich

Potassium phosphate monobasic

ACS reagent, ≥99.0%

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Agency

suitable for EPA 300
suitable for SM 5210

Assay

≥99.0%

form

powder or crystals

technique(s)

tissue culture: suitable

impurities

≤0.01% Insoluble matter

loss

≤0.2% loss on drying, 105°C

pH

4.1-4.5 (25 °C, 5%)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

bp

>450 °C

mp

252.6 °C (lit.)

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤0.002%
Na: ≤0.005%
heavy metals: ≤0.001% (by ICP)

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate monobasic (monopotassium orthophosphate) is a water soluble inorganic salt. Its standard enthalpy of formation has been reported to be -376.1kcalmol-1. It acts a starting material in the synthesis of potassium metaphosphate.

Application

Potassium phosphate monobasic has been used in the preparation of phosphate buffer and phosphate buffered saline (PBS). It has also been used in the preparation of PEG1000 (poly ethylene glycol)/potassium phosphate aqueous two-phase systems (ATPS).

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Standard enthalpies of formation of monopotassium and dipotassium orthophosphate.
Luff BB and Reed RB.
Journal of Chemical and Engineering Data, 23(1), 60-62 (1978)
Skin wound healing model-excisional wounding and assessment of lesion area.
Moreira CF, et al.
Bio-protocol, 5(22) (2015)
The solubility and reversion of calcium and potassium metaphosphates.
Volkerding CC and Bradfield R.
Soil Science Society of America Journal. Soil Science Society of America, 8(C), 159-166 (1944)
Estimation of Wound Tissue Neutrophil and Macrophage Accumulation by Measuring Myeloperoxidase (MPO) and N-Acetyl-?-D-glucosaminidase (NAG) Activities.
Cassini-Vieira P, et al.
Bio-protocol, 5(22) (2015)
Jacek Zielonka et al.
Free radical biology & medicine, 46(3), 329-338 (2008-11-26)
Methods for the detection and quantitation of hydroethidine (HE) and its oxidation products by HPLC analysis are described. Synthetic methods for preparation of authentic standards (2-hydroxyethidium and diethidium) are provided. Potential applications of the HPLC methods to chemical and biological

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service