Merck
  • Screening for anabolic steroids in urine of forensic cases using fully automated solid phase extraction and LC-MS-MS.
Screening for anabolic steroids in urine of forensic cases using fully automated solid phase extraction and LC-MS-MS.
Journal of analytical toxicology (2014-08-12)
David W Andersen, Kristian Linnet
ABSTRACT
A screening method for 18 frequently measured exogenous anabolic steroids and the testosterone/epitestosterone (T/E) ratio in forensic cases has been developed and validated. The method involves a fully automated sample preparation including enzyme treatment, addition of internal standards and solid phase extraction followed by analysis by liquid chromatography-tandem mass spectrometry (LC-MS-MS) using electrospray ionization with adduct formation for two compounds. Urine samples from 580 forensic cases were analyzed to determine the T/E ratio and occurrence of exogenous anabolic steroids. Extraction recoveries ranged from 77 to 95%, matrix effects from 48 to 78%, overall process efficiencies from 40 to 54% and the lower limit of identification ranged from 2 to 40 ng/mL. In the 580 urine samples analyzed from routine forensic cases, 17 (2.9%) were found positive for one or more anabolic steroids. Only seven different steroids including testosterone were found in the material, suggesting that only a small number of common steroids are likely to occur in a forensic context. The steroids were often in high concentrations (>100 ng/mL), and a combination of steroids and/or other drugs of abuse were seen in the majority of cases. The method presented serves as a fast and automated screening procedure, proving the suitability of LC-MS-MS for analyzing anabolic steroids.
Product Description
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%