Merck
  • Putative biomarker for phospholipid accumulation in cultured cells treated with phospholipidosis-inducing drugs: alteration of the phosphatidylinositol composition detected using high-performance liquid chromatography-tandem mass spectrometry.
Putative biomarker for phospholipid accumulation in cultured cells treated with phospholipidosis-inducing drugs: alteration of the phosphatidylinositol composition detected using high-performance liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-03)
Ryohei Hamaguchi, Toshiko Tanimoto, Yukihiro Kuroda
ABSTRACT
We developed a high-performance liquid chromatography-tandem mass spectrometric method for phospholipid biomarker discovery and applied it to a cell-based assay system for the screening of phospholipidosis-inducing drugs. We studied the compositions of phospholipid molecules exceeding 100 species in cultured cells and found a characteristic alteration in the composition by treatment with cationic amphiphilic drugs possessing phospholipidosis-inducing potency. The compositions of phosphatidylinositol in RAW264 cells were significantly affected by the drug treatment. Similar alterations were also found in THP-1 cells. These phenomena were not observed when cells were treated with warfarin, which does not have phospholipidosis-inducing potency. Structural analysis of the altered phosphatidylinositols by a product ion scan revealed the presence of certain fatty acyl chains. Based on our findings, we proposed a prediction parameter (PP) for phospholipid accumulation calculated from the relative compositions of phosphatidylinositol species. As the dosage of imipramine (a cationic amphiphilic drug) increased, both the PP and cellular phospholipid content increased. Our results suggest that PP has potency as a biomarker for phospholipid accumulation in cells treated with drugs.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Imipramine hydrochloride, ≥99% (TLC)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Tamoxifen citrate salt, ≥99%
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
Supelco
Imipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Tamoxifen citrate for performance test, European Pharmacopoeia (EP) Reference Standard
Imipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
USP
Imipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Warfarin Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Warfarin sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol