Skip to Content
Merck
All Photos(1)

Key Documents

392685

Sigma-Aldrich

Ammonia solution

2.0 M in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

form

liquid

Quality Level

concentration

1.90-2.20 M (with NaOH, titration)
2.0 M in ethanol

bp

60 °C

density

0.785 g/mL at 25 °C

application(s)

microbiology

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a solution of 2M ammonia in ethanol. Ammonia is a colorless gas with a characteristic strong odor. It is highly soluble in water, alcohol, chloroform and ether. Ammonia molecule is pyramidal in shape with nitrogen in the center and the three hydrogen atoms along the three vertices and the lone pair of electrons on the fourth vertex. Ammonia solution is commonly used as a base.

Application

Ammonia solution (in 2M ethanol) was used as an ammonia source to study the role of ammonia in causing metabolic changes in bacteria.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biogenic ammonia modifies antibiotic resistance at a distance in physically separated bacteria.
Bernier SP, et al.
Molecular Microbiology, 81(3), 705-716 (2011)
Ledgard J.
The Preparatory Manual of Explosives., 41-41 (2007)
Eagleson M.
Concise Encyclopedia Chemistry, 65-66 (1994)
Patrick M Erwin et al.
The ISME journal, 8(3), 575-588 (2013-10-25)
Ascidians are ecologically important components of marine ecosystems yet the ascidian microbiota remains largely unexplored beyond a few model species. We used 16S rRNA gene tag pyrosequencing to provide a comprehensive characterization of microbial symbionts in the tunic of 42
Olivier Braissant et al.
Journal of inherited metabolic disease, 36(4), 595-612 (2012-10-31)
Hyperammonemia can be caused by various acquired or inherited disorders such as urea cycle defects. The brain is much more susceptible to the deleterious effects of ammonium in childhood than in adulthood. Hyperammonemia provokes irreversible damage to the developing central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service