Merck
  • Lactoferrin-modified poly(ethylene glycol)-grafted BSA nanoparticles as a dual-targeting carrier for treating brain gliomas.
Lactoferrin-modified poly(ethylene glycol)-grafted BSA nanoparticles as a dual-targeting carrier for treating brain gliomas.
Molecular pharmaceutics (2014-05-02)
Zhigui Su, Lei Xing, Yinan Chen, Yurui Xu, Feifei Yang, Can Zhang, Qineng Ping, Yanyu Xiao
ABSTRACT
In this study, a dual-targeting drug delivery system based on bovine serum albumin nanoparticles (BSA-NPs) modified with both lactoferrin (Lf) and mPEG2000 loading doxorubicin (DOX) was designed, and its blood-brain barrier (BBB) penetration and brain glioma cells targeting properties were explored. BSA-NPs were prepared by a desolvation technique, and mPEG2000 was incorporated onto the surface of BSA-NPs by reacting with the free amino-group of BSA to form mPEG2000-modified BSA-NPs (P2000-NPs). Finally, Lf-modified P2000-NPs (Lf-NPs) was obtained by absorbing Lf onto the surface of P2000-NPs via the positive and negative charges interaction at physiological pH. Three levels of mPEG2000 and Lf-modified NPs were prepared and characterized, respectively. The uptake and potential cytotoxicity of different DOX preparations in vitro by the primary brain capillary endothelial cells (BCECs) and glioma cells (C6) were investigated. The dual-targeting effects were studied on the BBB model in vitro, BCECs/C6 glioma coculture model in vitro, and C6 glioma-bearing rats in vivo, respectively. The results exhibited that, with the increase of the amount of both mPEG2000 and Lf, the particle size of NPs increased and its zeta potential decreased. The in vivo pharmacokinetics study in healthy rats exhibited that P2000-NPs with a high level of mPEG2000 (P2000H-NPs) had longer circulation time in vivo. Compared to other NPs, Lf-NPs with high level of both Lf and mPEG2000 (LfH-NPs) showed the strongest cytotoxicity and the highest effectiveness in the uptake both in BCECs and C6 as well as improved the dual-targeting effects. Body distribution of DOX in different formulations revealed that LfH-NPs could significantly increase the accumulation of DOX in the brain, especially at 2 h postinjection (P < 0.05). In conclusion, Lf-NPs were a prospective dual-targeting drug delivery system for effective targeting therapy of brain gliomas.
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
4-Nitrophenyl chloroformate, 96%
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl ether, ≥99%
Sigma-Aldrich
Dimethyl ether, puriss., ≥99.9% (GC)