Skip to Content
Merck
All Photos(7)

Key Documents

793582

Sigma-Aldrich

Potassium iodide

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
grade:
anhydrous
form:
powder or crystals

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 745 °C)

product line

ReagentPlus®
Redi-Dri

Assay

99%

form

powder or crystals

quality

free-flowing

pH

~6.9 (20 °C, 50 g/L)

mp

681 °C (lit.)

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium iodide is an inorganic sodium salt. It is used as a source of iodide in organic synthesis. In the presence of oxidizing agents, it iodinates aromatic hydrocarbons and organothallium compounds. It is also used as a reagent for the cleavage of some ethers in the presence of acid.

Application

Potassium iodide can be used as a reducing agent for the reduction of graphene oxide in the presence of hydrochloric acid. It is also used as a dopant in the preparation of solid polymer electrolytes for energy devices.

Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders      
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs      
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents     
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danick Reynard et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(32), 7250-7257 (2020-04-09)
The MnIII /MnII redox couple with a standard potential of +1.51 V versus the standard hydrogen electrode (SHE) has attracted interest for the design of V/Mn redox flow batteries (RFBs). However, MnIII disproportionation leads to a loss of capacity, an increase
Pietro Santulli et al.
Human reproduction (Oxford, England), 30(1), 49-60 (2014-11-08)
Are protein oxidative stress markers [thiols, advanced oxidation protein products (AOPP), protein carbonyls and nitrates/nitrites] in perioperative peritoneal fluid higher in women with histologically proven endometriosis when compared with endometriosis-free controls? Protein oxidative stress markers are significantly increased in peritoneal
Dongjian Zhang et al.
Journal of drug targeting, 23(2), 180-190 (2014-10-21)
Necrosis-avid agents are a class of compounds that selectively accumulate in the necrotic tissues after systemic administration, which can be used for in vivo necrosis imaging and targeted therapies. In order to search for a necrosis-avid tracer agent with improved
Sungwook Choung et al.
Environmental science & technology, 48(16), 9684-9691 (2014-08-05)
Bentonite, which is used as an engineered barrier in geological repositories, is ineffective for sorbing anionic radionuclides because of its negatively charged surface. This study modified raw bentonite using a cationic surfactant (i.e., hexadecyltrimethylammonium [HDTMA]-Br) to improve its sorption capability
Ilzira A Minigalieva et al.
International journal of molecular sciences, 16(9), 22555-22583 (2015-09-24)
Stable suspensions of NiO and Mn₃O₄ nanoparticles (NPs) with a mean (±s.d.) diameter of 16.7±8.2 and 18.4±5.4 nm, respectively, purposefully prepared by laser ablation of 99.99% pure nickel or manganese in de-ionized water, were repeatedly injected intraperitoneally (IP) to rats

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service