Merck
  • The murine serotonin syndrome - evaluation of responses to 5-HT-enhancing drugs in NMRI mice.
The murine serotonin syndrome - evaluation of responses to 5-HT-enhancing drugs in NMRI mice.
Behavioural brain research (2014-05-02)
Robert Haberzettl, Heidrun Fink, Bettina Bert
ABSTRACT
In humans, the ingestion of the combination of two or more serotonin (5-HT)-enhancing drugs but also of a single drug in overdose can induce serious adverse effects, which are characteristics of the serotonin syndrome (SS). In mice, acute administration of direct and indirect 5-HT agonists also leads to behavioral and autonomic responses, but in literature different responses are thought to be essential. In order to detect common behavioral SS responses induced by 5-HT-enhancing drugs with different mechanisms of action, we investigated the effects of the 5-HT precursor 5-hydroxy-l-tryptophan (5-HTP), the selective serotonin reuptake inhibitor (SSRI) fluoxetine (FLX), and the monoaminooxidase (MAO) inhibitor tranylcypromine (TCP) in male NMRI mice. The drugs were administered alone or in combination to investigate additive effects or drug potentiation. Moreover, we compared the 5-HT responses to the effects induced by the dopamine, noradrenaline, and cholinergic agonists, apomorphine (APO), atomoxetine (ATO), and oxotremorine (OXO). Our results show that the studied 5-HT-enhancing drugs induced a different number of concomitant responses. The following five responses consistently and dose-dependently occurred in NMRI mice: flat body posture, hindlimb abduction, piloerection, tremor, and decreased rearings. Like in humans, the combination of 5-HT-enhancing drugs leads to a potentiation of drug effects. With the exception of flat body posture the responses are not specific for serotonergic hyperactivity. The findings demonstrate that the SS in NMRI mice is a suitable animal model for preclinical research, if it is taken into account that the spectrum of typical responses to 5-HT enhancing drugs may differ depending on drug and mouse strain and that some responses might be evoked by activation of other transmission systems, too.
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride, 97%
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
3,5,6-Trichloro-2-pyridinol, PESTANAL®, analytical standard
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
5-Hydroxy-L-tryptophan, 98% (calc. on dried substance)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%