Merck
  • A next-generation genetically attenuated Plasmodium falciparum parasite created by triple gene deletion.
A next-generation genetically attenuated Plasmodium falciparum parasite created by triple gene deletion.
Molecular therapy : the journal of the American Society of Gene Therapy (2014-05-16)
Sebastian A Mikolajczak, Viswanathan Lakshmanan, Matthew Fishbaugher, Nelly Camargo, Anke Harupa, Alexis Kaushansky, Alyse N Douglass, Michael Baldwin, Julie Healer, Matthew O'Neill, Thuan Phuong, Alan Cowman, Stefan H I Kappe
ABSTRACT
Immunization with live-attenuated Plasmodium sporozoites completely protects against malaria infection. Genetic engineering offers a versatile platform to create live-attenuated sporozoite vaccine candidates. We previously generated a genetically attenuated parasite (GAP) by deleting the P52 and P36 genes in the NF54 wild-type (WT) strain of Plasmodium falciparum (Pf p52(-)/p36(-) GAP). Preclinical assessment of p52(-)/p36(-) GAP in a humanized mouse model indicated an early and severe liver stage growth defect. However, human exposure to >200 Pf p52(-)/p36(-) GAP-infected mosquito bites in a safety trial resulted in peripheral parasitemia in one of six volunteers, revealing that this GAP was incompletely attenuated. We have now created a triple gene deleted GAP by additionally removing the SAP1 gene (Pf p52(-)/p36(-)/sap1(-) GAP) and employed flippase (FLP)/flippase recognition target (FRT) recombination for drug selectable marker cassette removal. This next-generation GAP was indistinguishable from WT parasites in blood stage and mosquito stage development. Using an improved humanized mouse model transplanted with human hepatocytes and human red blood cells, we show that despite a high-dose sporozoite challenge, Pf p52(-)/p36(-)/sap1(-) GAP did not transition to blood stage infection and appeared to be completely attenuated. Thus, clinical testing of Pf p52(-)/p36(-)/sap1(-) GAP assessing safety, immunogenicity, and efficacy against sporozoite challenge is warranted.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Flucytosine, European Pharmacopoeia (EP) Reference Standard
Flucytosine for system suitability, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog