Merck
  • Determination of chlorobenzenes in water samples by solid-phase disk extraction and gas chromatography-electron capture detection.
Determination of chlorobenzenes in water samples by solid-phase disk extraction and gas chromatography-electron capture detection.
Journal of chromatographic science (2013-05-07)
Hongmei Hu, Yuanming Guo, Xiumei Sun, Xuechang Chen, Xiaoning Zhang, Qin Liu, Chunxiu Xu
ABSTRACT
A simple, rapid, sensitive and high throughput method is described, based on solid-phase disk extraction (SPDE) and gas chromatography-electron capture detection, for the determination of chlorobenzens (CBs) in water samples. The proposed SPDE sample pretreatment method was initially optimized and the optimum experimental conditions were found to be as follows: 500 mL water sample (pH 2.5) extracted and enriched by an Empore 3-stn C18 (octadecyl) SPE disk at flow rate of 5 to 50 mL/min, eluted by 5 mL of acetone and 3 × 5 mL of methylene dichloride. The linearity of the method ranged from 0.02 to 0.4 µg/L for dichlorobenzene isomers, 0.0022-0.044 µg/L for trichlorobenzene isomers, 0.005-0.01 µg/L for tetrachlorobenzene isomers and 0.00025 to 0.005 µg/L for pentachlorobenzenes and hexachlorobenzenes, with correlation coefficients ranging between 0.9991 and 0.9999. The limits of detection were in the low ng/L level, ranging between 0.05 and 4 ng/L. The recoveries of spiked CBs with the external calibration method at different concentration levels in deionized/distilled water, tap water and sea water samples were 99-115, 91-106% and 96-110%, respectively, and with relative standard deviations of 4.5-7.6, 4.2-6.8 and 3.6-6.6% (n = 5), respectively. It is concluded that this method can successfully be applied for the determination of CBs in deionized/distilled water, tap water and sea water samples.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
1,3,5-Trichlorobenzene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
1,2,4-Trichlorobenzene, anhydrous, ≥99%
Supelco
1,2,4-Trichlorobenzene, PESTANAL®, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
1,3,5-Trichlorobenzene, 99%
Sigma-Aldrich
1,2,3-Trichlorobenzene, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%