Merck
  • Design, synthesis and in vitro antitumor activity of novel N-substituted-4-phenyl/benzylphthalazin-1-ones.
Design, synthesis and in vitro antitumor activity of novel N-substituted-4-phenyl/benzylphthalazin-1-ones.
European journal of medicinal chemistry (2014-12-03)
Wagdy M Eldehna, Hany S Ibrahim, Hatem A Abdel-Aziz, Noha N Farrag, Mohieldin M Youssef
ABSTRACT
A novel series of N-substituted-4-phenylphthalazin-1-ones 14a-g bearing different anilines at the N-2 of phthalazin-1-one scaffold via acetyl-flexible linker was designed and synthesized for the development of potential anticancer agents. Compounds 19a-g were synthesized by insertion of methylene (CH2) bridge at C4-position of 14a-g to provide a flexibility for the phenyl group. The newly synthesized compounds 14a-g and 19a-g were evaluated for their anti-proliferative activity against three human tumor cell lines HepG2 hepatocellular carcinoma, HT-29 colon cancer and MCF-7 breast cancer. In particular, HepG2 and HT-29 cancer cell lines were more susceptible to the synthesized derivatives. Compound 19d (IC50 = 1.2 ± 0.09 μM) was found to be the most potent derivative against HepG2 as it was 2.9 times more active than doxorubicin (IC50 = 3.45 ± 0.54) and sorafenib (IC50 = 3.5 ± 1.04 μM). Compounds 14e, 14g, 19d and 19g with IC50 = (3.29 ± 0.45), (3.50 ± 0.846), (1.20 ± 0.09) and (3.52 ± 0.70) μM, respectively, were found to be active candidates against HepG2 cancer cells. Compounds 14e, 14g, 19d and 19g were able to induce apoptosis in HepG2, this was assured by; the significant increase in the percentage of annexin V-FITC-positive apoptotic cells (UR + LR), the down-regulation of the anti-apoptotic protein Bcl-2 and the up-regulation of the pro-apoptotic protein Bax, in addition to boosting caspase-3 levels. Moreover, cytotoxicity evaluation of the newly synthesized compounds in HT-29 revealed that compounds 14e, 14f, 19e and 19f (IC50 = 3.05 ± 0.78, 4.02 ± 1.18, 3.68 ± 0.79 and 2.98 ± 0.47 μM, respectively) were more potent than doxorubicin (IC50 = 7.70 ± 1.78 μM).
Sigma-Aldrich
Ethanol, anhydrous, denatured
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
4-Phenyl-1-(2H)-phthalazinone, 97%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)