Merck
  • Effect of processing conditions on the organosulfides of shallot (Allium cepa L. Aggregatum group).
Effect of processing conditions on the organosulfides of shallot (Allium cepa L. Aggregatum group).
Journal of agricultural and food chemistry (2014-05-21)
Restituto Tocmo, Yi Lin, Dejian Huang
ABSTRACT
There is a growing account of the health benefits of H2S as an endogenous cell-signaling molecule. H2S from organic polysulfides, in particular, is increasingly gaining attention for their beneficial effects to cardiovascular health. Here, we studied shallot as a potential dietary source of organic polysulfides and examined the effects of processing conditions on its polysulfide profiles. Boiling, autoclaving, and freeze-drying were tested on whole and crushed shallot bulbs, analyzing their effect on the yield of organosulfides. Seventeen organosulfides, including disulfides, trisulfides, and cyclic polysulfides, were identified. Significant differences in the quantitative and qualitative profiles of organosulfides in the hydrodistilled and solvent extracted oils were observed. Freeze-drying retained the majority of the organosulfides, but the whole-autoclaved and whole-boiled shallots lost more than 95% of their organic polysulfides. Crushed-boiled and crushed-autoclaved shallot lost 76-80% of their organosulfides, likely due to the thermal sensitivity of these compounds. The organosulfide profiles are sensitive to the pH values of the processing media. In general, disulfides increased at basic pH (pH 9.0) while trisulfides and cyclic organosulfides are much higher at the acidic to neutral pH values (pH 3.0-5.0). Our results provide important information on the effects of processing conditions that are relevant for optimizing extraction of organosulfides from shallot for further studies evaluating their H2S-releasing activity.
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Methyl propyl disulfide, ≥95%, FG
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Dipropyl disulfide, 98%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Methyl propyl disulfide, 90%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl propyl trisulfide, 50%, FG
Supelco
Dimethyl trisulfide, analytical standard
Sigma-Aldrich
Dimethyl disulfide, ≥99%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dimethyl disulfide, ≥98%, FG