Skip to Content
Merck
All Photos(1)

Key Documents

71963

Sigma-Aldrich

Sodium sulfate

tested according to Ph. Eur., anhydrous

Synonym(s):

Natrii sulfas anhydricus

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

Assay

99.5% dry basis

form

crystalline

mp

884 °C (lit.)

solubility

water: soluble 185 g/L

density

2.68 g/mL at 25 °C (lit.)

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Betina Fejerskov et al.
ACS applied materials & interfaces, 4(9), 4981-4990 (2012-09-04)
In this work, we characterize physical hydrogels based on poly(vinyl alcohol), PVA, as intelligent biointerfaces for surface-mediated drug delivery. Specifically, we assemble microstructured (μS) surface adhered hydrogels via noncryogenic gelation of PVA, namely polymer coagulation using sodium sulfate (Na(2)SO(4)). We
Thomas Canam et al.
Plant science : an international journal of experimental plant biology, 198, 17-26 (2012-12-04)
Brassica carinata (Ethiopian mustard) has previously been identified as a potential crop species suitable for marginal land in the North American prairies due to its relatively high salt tolerance. Two genetically related B. carinata lines with brown-seeded (BS) and yellow-seeded
Clément Cartier et al.
Water research, 47(2), 661-671 (2012-11-24)
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the
Roosmarijn E Vandenbroucke et al.
Nature reviews. Drug discovery, 13(12), 904-927 (2014-11-08)
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases that form a family of 24 members in mammals. Evidence of the pathological roles of MMPs in various diseases, combined with their druggability, has made them attractive therapeutic targets. Initial drug discovery efforts focused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service