Merck
  • Interspecies comparison of hepatic metabolism of six newly synthesized retinoid X receptor agonistic compounds possessing a 6-[N-ethyl-N-(alkoxyisopropylphenyl)amino]nicotinic acid skeleton in rat and human liver microsomes.
Interspecies comparison of hepatic metabolism of six newly synthesized retinoid X receptor agonistic compounds possessing a 6-[N-ethyl-N-(alkoxyisopropylphenyl)amino]nicotinic acid skeleton in rat and human liver microsomes.
Drug development and industrial pharmacy (2013-06-21)
Yoshiki Murakami, Yasumasa Shimizu, Akemi Ogasawara, Satoshi Ueshima, Mariko Nakayama, Kohei Kawata, Hiroki Kakuta, Tetsuya Aiba
ABSTRACT
The hepatic metabolism of six compounds newly synthesized as retinoid X receptor agonists was characterized in rat and human liver microsomes to examine the relationship between their hepatic metabolism profiles and side chain structures, considering the interspecies difference. The compounds used have a 6-[N-ethyl-N-(3-alkoxy-4-isopropylphenyl)amino]nicotinic or 6-[N-ethyl-N-(4-alkoxy-3-isopropylphenyl)amino]nicotinic acid skeleton, in which the isopropoxy, isobutoxy or cyclopropylmethoxy group is employed for the alkoxy group. These compounds were incubated with the microsomes, and their Michaelis--Menten parameters were determined. The incubation study was also performed with various cytochrome P450 (CYP) inhibitors to examine their susceptibilities to the inhibitors. In addition, a molecular docking simulation was conducted to assess the compound's spatial configuration with the CYP isoform when necessary. The Michaelis--Menten parameters determined are comparable between rats and humans for the compounds having 3-isobutoxy, 4-isobutoxy, 4-isopropoxy and 4-cyclopropylmethoxy groups. However, it was indicated that all compounds except that having the 3-isobutoxy group are metabolized in a different manner between rats and humans. That is, the extent of the contribution of each CYP isozyme is different between those two species. A molecular docking simulation showed that the spatial configuration of the compound to be associated with CYP2D6 markedly changes depending on whether the isobutoxy group is situated at the 3- or 4-position. A slight difference in the side chain structures markedly alters the compound's metabolic profile, which amplifies the interspecies difference regarding the profile, increasing the difficulty in characterizing the profile in humans with the structural-property relationship and interspecies extrapolation.
Product Description
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Sigma-Aldrich
Ethanol, anhydrous, denatured
Supelco
Sulfadimethoxine, 98.0-102.0%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Sulfadimethoxine, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Quinidine, anhydrous
Supelco
8-Methoxypsoralen, analytical standard
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Xanthotoxin, analytical standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material