Merck
  • Structural control of hierarchically-ordered TiO2 films by water for dye-sensitized solar cells.
Structural control of hierarchically-ordered TiO2 films by water for dye-sensitized solar cells.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-05-13)
Sung Hoon Ahn, Dong Jun Kim, Dong Kyu Roh, Won Seok Chi, Jong Hak Kim
ABSTRACT
A facile way of controlling the structure of TiO(2) by changing the amount of water to improve the efficiency of dye-sensitized solar cells (DSSCs) is reported. Hierarchically ordered TiO(2) films with high porosity and good interconnectivity are synthesized in a well-defined morphological confinement arising from a one-step self-assembly of preformed TiO(2) (pre-TiO(2)) nanocrystals and a graft copolymer, namely poly(vinyl chloride)-g-poly(oxyethylene methacrylate). The polymer-solvent interactions in solution, which are tuned by the amount of water, are shown to be a decisive factor in determining TiO(2) morphology and device performance. Systematic control of wall and pore size is achieved and enables the bifunctionality of excellent light scattering properties and easy electron transport through the film. These properties are characterized by reflectance spectroscopy, incident photon-to-electron conversion efficiency, and electrochemical impedance spectroscopy analyses. The TiO(2) photoanode that is prepared with a higher water ratio, [pre-TiO(2)]:[H(2)O]=1:0.3, shows a larger surface area, greater light scattering, and better electron transport, which result in a high efficiency (7.7 %) DSSC with a solid polymerized ionic liquid. This efficiency is much greater than that of commercially available TiO(2) paste (4.0 %).
Sigma-Aldrich
Titanium(IV) isopropoxide, packaged for use in deposition systems
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Titanium(IV) isopropoxide, ≥97.0%
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Titanium(IV) isopropoxide, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Titanium(IV) isopropoxide, 97%
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Chloroplatinic acid hexahydrate, BioXtra
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Chloroplatinic acid hexahydrate, powder and chunks
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroplatinic acid hexahydrate, ACS reagent, ≥37.50% Pt basis
Sigma-Aldrich
1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%
Sigma-Aldrich
Ethylene glycol 5 M solution