Skip to Content
Merck
All Photos(1)

Key Documents

P5775

Sigma-Aldrich

Chloroplatinic acid hexahydrate

powder and chunks

Synonym(s):

Hexachloroplatinic acid hexahydrate, Hydrogen hexachloroplatinate(IV) hexahydrate, Platinic chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2PtCl6 · 6H2O
CAS Number:
Molecular Weight:
517.90
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.21

Pricing and availability is not currently available. Contact Technical Service

form

powder and chunks

Quality Level

concentration

≥37.5% Pt (after Reduction with Mg, gravimetric)

color

orange

solubility

water: 100 mg/mL, clear to slightly hazy, yellow to orange

storage temp.

room temp

SMILES string

O.O.O.O.O.O.Cl.Cl.Cl[Pt](Cl)(Cl)Cl

InChI

1S/6ClH.6H2O.Pt/h6*1H;6*1H2;/q;;;;;;;;;;;;+4/p-4

InChI key

PIJUVEPNGATXOD-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Han et al.
Scientific reports, 4, 6414-6414 (2014-09-19)
Noble metal nanoparticles with hollow interiors and customizable shell compositions have immense potential for catalysis. Herein, we present an unique structure transformation phenomenon for the fabrication of alloy Cu₃Pt nanoframes with polyhedral morphology. This strategy starts with the preparation of
Sung Hoon Ahn et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(9), 1841-1848 (2014-05-13)
A facile way of controlling the structure of TiO(2) by changing the amount of water to improve the efficiency of dye-sensitized solar cells (DSSCs) is reported. Hierarchically ordered TiO(2) films with high porosity and good interconnectivity are synthesized in a
Rui-Xiang Wang et al.
Nanoscale, 6(24), 14999-15007 (2014-11-05)
A novel nanostructured catalyst of platinum nanoparticles supported on 5,10,15,20-tetrakis(1-methyl-4-pyridinio)porphyrin tetra(p-toluenesulfonate) (TMPyP) functionalized graphene (TMPyP-graphene) is synthesized by the hydrothermal polyol process. The as-synthesized nanocomposites are characterized by Fourier transform infrared (FTIR) spectroscopy, UV-vis absorption spectroscopy, Raman spectroscopy, X-ray diffraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service