Merck
  • Design, synthesis, and bioevaluation of paeonol derivatives as potential anti-HBV agents.
Design, synthesis, and bioevaluation of paeonol derivatives as potential anti-HBV agents.
European journal of medicinal chemistry (2014-12-03)
Tsurng-Juhn Huang, Hong Chuang, Yu-Chuan Liang, Hui-Hsien Lin, Jia-Cherng Horng, Yu-Cheng Kuo, Chia-Wen Chen, Fu-Yuan Tsai, Shih-Chieh Yen, Shih-Ching Chou, Ming-Hua Hsu
ABSTRACT
Hepatitis B virus (HBV) is a causative reagent that frequently causes progressive liver diseases, leading to the development of acute, chronic hepatitis, cirrhosis, and eventually hepatocellular carcinoma (HCC). Despite several antiviral drugs including interferon-α and nucleotide derivatives are approved for clinical treatment for HBV, critical issues remain unresolved, e.g., low-to-moderate efficacy, adverse side effects, and resistant strains. In this study, novel Paeonol-phenylsulfonyl derivatives were synthesized and their antiviral effect against HBV was evaluated. The experimental results indicated that these compounds process significant antiviral potential, including the inhibition of viral antigen expression and secretion, and the suppression of HBV viral DNA replication. Among compounds synthesized in this research, compound 2-acetyl-5-methoxyphenyl 4-methoxybenzenesulfonate (7f) had the most potent inhibitory activity with IC50 value of 0.36 μM, and high selectivity index, SI (TC50/IC50) 47.75; which exhibited an apparent inhibition effect on viral gene expression and viral propagation in cell culture model. So, we believe our compounds could serve as reservoir for antiviral drug development.
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Hexane, analytical standard
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
4-Bromobenzenesulfonyl chloride, 98%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent