Skip to Content
Merck
All Photos(2)

Key Documents

590681

Sigma-Aldrich

Potassium carbonate

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Carbonate of potash, Dipotassium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder
solubility:
ethanol: insoluble(lit.)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

891 °C (lit.)

solubility

ethanol: insoluble(lit.)

density

2.43 g/mL at 25 °C

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium carbonate may be used as a base in the synthesis of phytochemical-based oryzanol modified polystyrene from 4-chloromethylstyrene and γ-oryzanol in order to study their liquid crystal (LC) orientation property.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vertical liquid crystal orientation of phytochemical-based oryzanol modified polystyrene.
Kim T, et al.
Royal Society of Chemistry Advances, 8(3), 1569-1575 (2018)
S V Hartimath et al.
Molecular metabolism, 25, 154-158 (2019-05-21)
The browning of white adipose tissue (WAT) into beige has been proposed as a strategy to enhance energy expenditure to combat the growing epidemic of obesity. Research into browning strategies are hampered by the lack of sensitive, translatable, imaging tools
K Y Foo et al.
Bioresource technology, 102(20), 9814-9817 (2011-08-30)
Rice husk (RH), an abundant by-product of rice milling, was used for the preparation of activated carbon (RHAC) via KOH and K(2)CO(3) chemical activation. The activation process was performed at the microwave input power of 600 W for 7 min.
Carlos Arroniz et al.
Organic letters, 15(4), 910-913 (2013-02-05)
ortho-Arylation of ortho-substituted benzoic acids is a challenging process due to the tendency of the reaction products toward Pd-catalyzed protodecarboxylation. A simple method for preventing decarboxylation in sterically hindered benzoic acids is reported. The method described represents a reliable and
K Y Foo et al.
Bioresource technology, 102(20), 9794-9799 (2011-08-31)
Sunflower seed oil residue, a by-product of sunflower seed oil refining, was utilized as a feedstock for preparation of activated carbon (SSHAC) via microwave induced K(2)CO(3) chemical activation. SSHAC was characterized by Fourier transform infrared spectroscopy, nitrogen adsorption-desorption and elemental

Articles

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service