Merck
  • Quantification of piperacillin, tazobactam, cefepime, meropenem, ciprofloxacin and linezolid in serum using an isotope dilution UHPLC-MS/MS method with semi-automated sample preparation.
Quantification of piperacillin, tazobactam, cefepime, meropenem, ciprofloxacin and linezolid in serum using an isotope dilution UHPLC-MS/MS method with semi-automated sample preparation.
Clinical chemistry and laboratory medicine (2014-10-11)
Johannes Zander, Barbara Maier, Anna Suhr, Michael Zoller, Lorenz Frey, Daniel Teupser, Michael Vogeser
ABSTRACT
Recent studies have demonstrated highly variable blood concentrations of piperacillin, tazobactam, cefepime, meropenem, ciprofloxacin and linezolid in critically ill patients with a high incidence of sub-therapeutic levels. Consequently, therapeutic drug monitoring (TDM) of these antibiotics has to be considered, requiring robust and reliable routine analytical methods. The aim of the present work was to develop and validate a multi-analyte ultra high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) method for the simultaneous quantification of the above mentioned antibiotics. Sample preparation included a manual protein precipitation step followed by two-dimensional ultra high performance liquid chromatography (2D-UHPLC). Corresponding stable isotope-labeled substances were used as internal standards for all of the analytes, with the exception of tazobactam. The injected sample volume was 7 μL. The run time was 5.0 min. Inaccuracy was ≤8% and imprecision coefficient of variation (CV) was <9% for all analytes. Only minor matrix effects and negligible carry-over was observed. The method was found to be robust during the validation period. We were able to develop a reliable 2D-UHPLC-MS/MS method addressing analytes with highly heterogeneous physico-chemical properties. The novel assay may be an efficient tool for an optimized process workflow in clinical laboratories for important antibiotics in regards to TDM.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Piperacillin sodium salt, penicillin analog
Supelco
Piperacillin sodium salt, analytical standard
Sigma-Aldrich
Tazobactam sodium salt, β-lactamase inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Meropenem trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Meropenem trihydrate, ≥98% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Piperacillin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%