Skip to Content
Merck
All Photos(2)

Key Documents

T2820

Sigma-Aldrich

Tazobactam sodium salt

β-lactamase inhibitor

Synonym(s):

CL-298741, YTR-830H, [2S-(2α, 3β, 5α)]-3-Methyl-7-oxo-3(1H,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N4NaO5S
CAS Number:
Molecular Weight:
322.27
MDL number:
UNSPSC Code:
51282127
PubChem Substance ID:
NACRES:
NA.85

Quality Level

form

powder or crystals

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

SMILES string

[Na+].C[C@]2(Cn1ccnn1)[C@@H](N3[C@@H](CC3=O)S2(=O)=O)C([O-])=O

InChI

1S/C10H12N4O5S.Na/c1-10(5-13-3-2-11-12-13)8(9(16)17)14-6(15)4-7(14)20(10,18)19;/h2-3,7-8H,4-5H2,1H3,(H,16,17);/q;+1/p-1/t7-,8+,10+;/m1./s1

InChI key

RFMIKMMOLPNEDG-QVUDESDKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Tazobactam is an antibacterial penicillin derivative which inhibits the action of bacterial β-lactamases. It is used in combination with piperacillin and other β-lactam antibiotics to broaden their spectrum and enhance their effect. It is used for bacterial β-lactamase and penicillinase inhibition studies.

Biochem/physiol Actions

Tazobactam is a β -Lactamase inhibitor. When used with β-lactam antibiotics I, it enhances their effect.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro antimicrobial activity of piperacillin/tazobactam in comparison with other broad-spectrum β-lactams. .
Roland, R. K., et al.
The Journal of Infectious Diseases, 4, 226-226 (2000)
Gioia S Babini et al.
The Journal of antimicrobial chemotherapy, 51(3), 605-612 (2003-03-05)
MICs of piperacillin/tazobactam are conventionally determined by varying the concentration of piperacillin in the presence of a fixed 4 mg/L tazobactam. When tested in this way, the MIC distribution for Klebsiella isolates with extended-spectrum beta-lactamases (ESBLs) is strongly bimodal, such
R A Bonomo et al.
Biochimica et biophysica acta, 1547(2), 196-205 (2001-06-19)
The CMY-2 beta-lactamase, a plasmid determined class C cephalosporinase, was shown to be susceptible to inhibition by tazobactam (K(i)=40 microM). The reaction product(s) of CMY-2 beta-lactamase with the beta-lactamase inhibitor tazobactam were analyzed by electrospray ionization/mass spectrometry (ESI/MS) to characterize
Craig Knox et al.
Nucleic acids research, 39(Database issue), D1035-D1041 (2010-11-10)
DrugBank (http://www.drugbank.ca) is a richly annotated database of drug and drug target information. It contains extensive data on the nomenclature, ontology, chemistry, structure, function, action, pharmacology, pharmacokinetics, metabolism and pharmaceutical properties of both small molecule and large molecule (biotech) drugs.
Johannes Zander et al.
Clinical chemistry and laboratory medicine, 53(5), 781-791 (2014-10-11)
Recent studies have demonstrated highly variable blood concentrations of piperacillin, tazobactam, cefepime, meropenem, ciprofloxacin and linezolid in critically ill patients with a high incidence of sub-therapeutic levels. Consequently, therapeutic drug monitoring (TDM) of these antibiotics has to be considered, requiring

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service