Merck
  • Simplified matrix solid phase dispersion procedure for the determination of parabens and benzophenone-ultraviolet filters in human placental tissue samples.
Simplified matrix solid phase dispersion procedure for the determination of parabens and benzophenone-ultraviolet filters in human placental tissue samples.
Journal of chromatography. A (2014-12-03)
F Vela-Soria, I Rodríguez, O Ballesteros, A Zafra-Gómez, L Ballesteros, R Cela, A Navalón
ABSTRACT
In recent decades, the industrial development has resulted in the appearance of a large amount of new chemicals that are able to produce disorders in the human endocrine system. These substances, so-called endocrine disrupting chemicals (EDCs), include many families of compounds, such as parabens and benzophenone-UV filters. Taking into account the demonstrated biological activity of these compounds, it is necessary to develop new analytical procedures to assess the exposure in order to establish, in an accurate way, relationships between EDCs and harmful health effects in population. In the present work, a new method based on a simplified sample treatment by matrix solid phase dispersion (MSPD) followed by ultrahigh performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) analysis, is validated for the determination of four parabens (methyl-, ethyl-, propyl- and butylparaben) and six benzophenone-UV filters (benzophenone-1, benzophenone-2, benzophenone-3, benzophenone-6, benzophenone-8 and 4-hydroxybenzophenone) in human placental tissue samples. The extraction parameters were accurately optimized using multivariate optimization strategies. Ethylparaben ring-13C6 and benzophenone-d10 were used as surrogates. The found limits of quantification ranged from 0.2 to 0.4 ng g(-1) and inter-day variability (evaluated as relative standard deviation) ranged from 5.4% to 12.8%. The method was validated using matrix-matched standard calibration followed by a recovery assay with spiked samples. Recovery rates ranged from 96% to 104%. The method was satisfactorily applied for the determination of compounds in human placental tissue samples collected at the moment of delivery from 10 randomly selected women.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Ethyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
USP
Ethylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%