Merck
  • A comprehensive platform to investigate protein-metal ion interactions by affinity capillary electrophoresis.
A comprehensive platform to investigate protein-metal ion interactions by affinity capillary electrophoresis.
Journal of pharmaceutical and biomedical analysis (2015-02-02)
Hassan A Alhazmi, Markus Nachbar, Hassan M Albishri, Deia Abd El-Hady, Sabine Redweik, Sami El Deeb, Hermann Wätzig
ABSTRACT
In this work, the behavior of several metal ions with different globular proteins was investigated by affinity capillary electrophoresis. Screening was conducted by applying a proper rinsing protocol developed by our group. The use of 0.1M EDTA in the rinsing solution successfully desorbs metal ions from the capillary wall. The mobility ratio was used to evaluate the precision of the method. Excellent precision for repeated runs was achieved for different protein metal ion interactions (RSD% of 0.05-1.0%). Run times were less than 6 min for all of the investigated interactions. The method has been successfully applied for the interaction study of Li(+), Na(+), Mg(2+), Ca(2+), Ba(2+), Al(3+), Ga(3+), La(3+), Pd(2+), Ir(3+), Ru(3+), Rh(3+), Pt(2+), Pt(4+), Os(3+), Au(3+), Au(+), Ag(+), Cu(1+), Cu(2+), Fe(2+), Fe(3+), Co(2+), Ni(2+), Cr(3+), V(3+), MoO4(2-) and SeO3(2-) with bovine serum albumin, ovalbumin, β-lactoglobulin and myoglobin. Different interaction values were obtained for most of the tested metal ions even for that in the same metal group. Results were discussed and compared in view of metal and semimetal group's interaction behavior with the tested proteins. The calculated normalized difference of mobility ratios for each protein-metal ion interaction and its sign (positive and negative) has been successfully used to detect the interaction and estimate further coordination of the bound metal ion, respectively. The comprehensive platform summarizes all the obtained interaction results, and is valuable for any future protein-metal ion investigation.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Malonic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Acetanilide, purified by sublimation, ≥99.9%
Sigma-Aldrich
Acetanilide, zone-refined, purified by sublimation, ≥99.95%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetanilide, 99%
Sigma-Aldrich
Acetanilide, puriss. p.a., ≥99.5% (CHN)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Supelco
Acetaminophen Related Compound D, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetanilide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetanilide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard