Merck
  • Comparison of different urine pretreatments for biological monitoring of pyrethroid insecticides.
Comparison of different urine pretreatments for biological monitoring of pyrethroid insecticides.
Journal of analytical toxicology (2014-12-30)
Hiroki Toshima, Jun Yoshinaga, Hiroaki Shiraishi, Yuki Ito, Michihiro Kamijima, Jun Ueyama
ABSTRACT
During a cross-validation practice of urinary analysis of pyrethroid insecticide metabolite (3-phenoxybenzoic acid, 3-PBA) by two laboratories (Lab A and Lab B), difference in 3-PBA concentration was noticed. The analytical methods used in the exercise were enzymatic deconjugation (glucuronidase/sulfatase) followed by solid phase extraction and isotope dilution LC-MS-MS determination (Lab A) and acidic deconjugation followed by liquid-liquid extraction and gas chromatography-mass spectrometry determination (Lab B). A significant difference was found for one of the five samples: lower value was obtained in Lab A. Use of acidic deconjugation in Lab A resulted in comparable value with the analytical result for this sample originally obtained in Lab B. The comparison was extended to 42 urine samples taken from Japanese males in Lab A by using the two different deconjugation methods and LC-MS-MS determination. Significantly lower value was obtained for enzymatic deconjugation in some of the 42 urine samples. The results suggested the presence of other conjugated species of 3-PBA than glucuronide and sulfate in human urine. Although the overall agreement between the values obtained by the deconjugation methods was fair, it appears that urine samples should be pretreated by acidic deconjugation for the analysis in biological monitoring of pyrethroid exposure.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
tert-Butyl methyl ether, analytical standard
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%