Merck
  • Following the azide-alkyne cycloaddition at the silica/solvent interface with sum frequency generation.
Following the azide-alkyne cycloaddition at the silica/solvent interface with sum frequency generation.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-05-08)
Zhiguo Li, Champika N Weeraman, Julianne M Gibbs-Davis
ABSTRACT
The Cu(I) -catalyzed 1,3-dipolar azide-alkyne cycloaddition (CuAAC) has arisen as one of the most useful chemical transformations for introducing complexity onto surfaces and materials owing to its functional-group tolerance and high yield. However, methods for monitoring such reactions in situ at the widely used silica/solvent interface are hampered by challenges associated with probing such buried interfaces. Using the surface-specific technique broadband sum frequency generation (SFG), we monitored the reaction of a benzyl azide monolayer in real time at the silica/methanol interface. A strong peak at 2096 cm(-1) assigned to the azides was observed for the first time by SFG. Using a cyano-substituted alkyne, the decrease of the azide peak and the increase of the cyano peak (2234 cm(-1) ) were probed simultaneously. From the kinetic analysis, the reaction order with respect to copper was determined to be 2.1, suggesting that CuAAC on the surface follows a similar mechanism as in solution.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss., suitable for, meets analytical specification of Ph. Eur., BP, USP,FCC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99-102%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1-Hexyne, 97%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))