Skip to Content
Merck
All Photos(4)

Key Documents

C7631

Sigma-Aldrich

Copper(II) sulfate pentahydrate

ReagentPlus®, ≥98.0%

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0%
form:
powder or crystals

vapor pressure

7.3 mmHg ( 25 °C)

Quality Level

product line

ReagentPlus®

Assay

≥98.0%

form

powder or crystals

pH

3.7-4.5 (25 °C, 50 g/L)

mp

110 °C (dec.) (lit.)

cation traces

Fe: ≤0.01%

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Copper(II) sulfate is an inorganic Lewis acid commonly used to promote acid?catalyzed organic reactions. It is used as a reagent for the synthesis of copper carbenoids. It can also act as an effective redox catalyst in combination with other mixed oxidizing systems.

Application


  • Determination of main fruits in adulterated nectars by ATR-FTIR spectroscopy combined with multivariate calibration and variable selection methods.: This study highlights the application of ATR-FTIR spectroscopy, a technique involving Copper(II) sulfate pentahydrate, in identifying fruit content in adulterated nectars, illustrating its relevance in food chemistry and quality control (de Souza SVC et al., 2018).

  • Measurement of electron paramagnetic resonance using terahertz time-domain spectroscopy.: This research demonstrates the use of terahertz time-domain spectroscopy, involving Copper(II) sulfate pentahydrate, to measure electron paramagnetic resonance, contributing to advancements in spectroscopic methodologies (Hangyo M et al., 2011).

  • Effects of Curcuma xanthorrhiza Extracts and Their Constituents on Phase II Drug-metabolizing Enzymes Activity.: Though indirectly related, this article explores the role of natural extracts on enzyme activity, providing a context where Copper(II) sulfate pentahydrate could be used as a comparative standard in bioanalytical settings (Ab Halim MR et al., 2016).


Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper(II) Sulfate
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2016)
Bárbara Acosta-Iborra et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(5), 6654-6674 (2020-03-31)
Angiogenesis, the main mechanism that allows vascular expansion for tissue regeneration or disease progression, is often triggered by an imbalance between oxygen consumption and demand. Here, by analyzing changes in the transcriptomic profile of endothelial cells (ECs) under hypoxia we
Yuan Chen et al.
Biochemical and biophysical research communications, 459(3), 529-533 (2015-03-10)
Apolipoprotein E (ApoE) genotypes are related to clinical presentations in patients with Wilson's disease, indicating that ApoE may play an important role in the disease. However, our understanding of the role of ApoE in Wilson's disease is limited. High copper
Gabriela M B Haverroth et al.
Ecotoxicology and environmental safety, 122, 440-447 (2015-09-20)
Copper is a heavy metal found at relatively high concentrations in surface waters around the world. Copper is a micronutrient at low concentrations and is essential to several organisms. At higher concentrations copper can become toxic, which reveal the importance
Bhagwan S Batule et al.
International journal of nanomedicine, 10 Spec Iss, 137-142 (2015-09-09)
We developed a simple but efficient method to synthesize protein-inorganic hybrid nanostructures with a flower-like shape (nanoflowers), which relies on sonication to facilitate the synthesis of the nanoflowers. With this technique, we synthesized nanoflowers containing laccase as a model protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service