Merck
  • Water/oil type microemulsion systems containing lidocaine hydrochloride: in vitro and in vivo evaluation.
Water/oil type microemulsion systems containing lidocaine hydrochloride: in vitro and in vivo evaluation.
Journal of microencapsulation (2014-04-05)
Ahmet Dogrul, Seyda Akkus Arslan, Figen Tirnaksiz
ABSTRACT
The purpose of this study was to develop a water/oil microemulsion containing lidocaine hydrochloride (4%) and to compare its local anaesthetic efficacy with commercial products. A pseudoternary diagram (Km:1/1 or 1/2) was constructed using lecithin/ethanol/oil/water. The droplet size, viscosity and release of the microemulsions were evaluated. Tail flick tests were conducted for in vivo effectiveness; the initiation time of effect, maximum effect, time to reach maximum effect, and relative efficacy were evaluated. The drug caused a significant increase in droplet size. The use of olive oil resulted in a decrease in the solubilisation parameter, as well as a reduction in the release. The droplet size and viscosity of the microemulsion composed of Miglyol/lecithin/ethanol/water/drug (Km:1/2) was lower than other microemulsions (8.38 nm, 6.9 mPa), and its release rate (1.61 mg/h) was higher. This system had a faster and more efficient anaesthetic effect than the other microemulsions and commercial products. Results indicate that a water/oil type microemulsion (Miglyol/lecithin/ethanol/water) has promising potential to increase the local anaesthetic effect.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, anhydrous, denatured
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dialysis tubing, high retention seamless cellulose tubing, avg. flat width 23 mm (0.9 in.), MWCO 14000, 99.99% retention