Merck
  • Morgana acts as a proto-oncogene through inhibition of a ROCK-PTEN pathway.
Morgana acts as a proto-oncogene through inhibition of a ROCK-PTEN pathway.
The Journal of pathology (2014-03-13)
Federica Fusella, Roberta Ferretti, Daniele Recupero, Stefania Rocca, Augusta Di Savino, Giusy Tornillo, Lorenzo Silengo, Emilia Turco, Sara Cabodi, Paolo Provero, Pier Paolo Pandolfi, Anna Sapino, Guido Tarone, Mara Brancaccio
ABSTRACT
Morgana/CHP-1 is a ubiquitously expressed protein able to inhibit ROCK II kinase activity. We have previously demonstrated that morgana haploinsufficiency leads to multiple centrosomes, genomic instability, and higher susceptibility to tumour development. While a large fraction of human cancers has shown morgana down-regulation, a small subset of tumours was shown to express high morgana levels. Here we demonstrate that high morgana expression in different breast cancer subtypes correlates with high tumour grade, mitosis number, and lymph node positivity. Moreover, morgana overexpression induces transformation in NIH-3T3 cells and strongly protects them from various apoptotic stimuli. From a mechanistic point of view, we demonstrate that morgana causes PTEN destabilization, by inhibiting ROCK activity, hence triggering the PI3K/AKT survival pathway. In turn, morgana down-regulation in breast cancer cells that express high morgana levels increases PTEN expression and leads to sensitization of cells to chemotherapy.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Chordc1
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone V284, purified from hybridoma cell culture
USP
Docetaxel, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
GSK690693, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human CHORDC1
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse