Skip to Content
Merck
All Photos(4)

Key Documents

H3160

Sigma-Aldrich

Hydrocortisone

meets USP testing specifications

Synonym(s):

11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Beilstein:
1354819
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

plant (wild yam)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

97.0-102.0% dry basis

form

powder

potency

0.004-0.005 μg/mL

mp

211-214 °C (lit.)

solubility

water: soluble 0.28 g/L at 25 °C

application(s)

pharmaceutical (small molecule)

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Exploring Microemulsion Systems for the Incorporation of Glucocorticoids into Bacterial Cellulose: This research presents a novel method for embedding hydrocortisone into bacterial cellulose using microemulsion systems, highlighting its potential for creating anti-inflammatory wound dressings, a significant advancement in pharmaceutical design and material science (Zahel et al., 2024).

  • Research progress in the treatment of chronic fatigue syndrome through interventions targeting the hypothalamus-pituitary-adrenal axis: This review discusses the role of hydrocortisone in treating chronic fatigue syndrome by modulating the hypothalamus-pituitary-adrenal axis, providing a comprehensive overview of its therapeutic applications in endocrinology (Zhang and Wang, 2024).

Biochem/physiol Actions

Primary glucocorticoid secreted by the adrenal cortex.
Primary glucocorticoid secreted by the adrenal cortex. It has three times the anti-inflammatory potency of corticosterone but much lower Na2+ retention potency.

Physical form

powder-RT; stock-frozen in working aliquots, avoid repeated freeze/thaw

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margriet A B Veldhorst et al.
The Journal of clinical endocrinology and metabolism, 99(1), 285-290 (2014-01-05)
Pathologically increased cortisol exposure induces obesity, but it is not known whether relatively high cortisol within the physiological range is related to childhood obesity. The aim of the study was to compare hair cortisol concentrations between obese and normal-weight children.
Lorenz Goette et al.
Psychoneuroendocrinology, 54, 115-123 (2015-02-24)
Social competition is a fundamental mechanism of evolution and plays a central role in structuring individual interactions and communities. Little is known about the factors that affect individuals' competitive success, particularly in humans. Key factors might include stress, a major
Florian Hardeveld et al.
Psychoneuroendocrinology, 50, 62-71 (2014-09-03)
Although HPA-axis activity has been studied extensively in relation to depression, there is no consensus whether HPA-axis parameters predicts major depressive disorder (MDD) recurrence. We investigated whether HPA-axis parameters (cortisol awakening response (CAR), the dexamethasone suppression test (DST) and evening
Rachel Yehuda et al.
The American journal of psychiatry, 171(8), 872-880 (2014-05-17)
Differential effects of maternal and paternal posttraumatic stress disorder (PTSD) have been observed in adult offspring of Holocaust survivors in both glucocorticoid receptor sensitivity and vulnerability to psychiatric disorder. The authors examined the relative influences of maternal and paternal PTSD
Marianne Oksnes et al.
The Journal of clinical endocrinology and metabolism, 99(5), 1665-1674 (2014-02-13)
Conventional glucocorticoid replacement therapy fails to mimic the physiological cortisol rhythm, which may have implications for morbidity and mortality in patients with Addison's disease. The objective of the study was to compare the effects of continuous sc hydrocortisone infusion (CSHI)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service