Merck
  • Comparison of smartphone ophthalmoscopy with slit-lamp biomicroscopy for grading diabetic retinopathy.
Comparison of smartphone ophthalmoscopy with slit-lamp biomicroscopy for grading diabetic retinopathy.
American journal of ophthalmology (2014-12-03)
Andrea Russo, Francesco Morescalchi, Ciro Costagliola, Luisa Delcassi, Francesco Semeraro
ABSTRACT
To assess the accuracy and reliability of smartphone ophthalmoscopy, we compared the ability of a smartphone ophthalmoscope with that of a slit-lamp biomicroscope to grade diabetic retinopathy (DR) in patients with diabetes mellitus (DM). Clinical-based, prospective, comparative instrument study. This comparative clinical study was performed in 120 outpatients (240 eyes) with type 1 or type 2 DM. After pupil dilation, the patients underwent smartphone ophthalmoscopy with the D-Eye device, followed by dilated retinal slit-lamp examination, to grade DR according to a 5-step scale. Overall exact agreement between the 2 methods was observed in 204 of 240 eyes (85%) (simple κ = 0.78; CI 0.71-0.84) and agreement within 1 step was observed in 232 eyes (96.7%). Compared to biomicroscopy, the sensitivity and specificity of smartphone ophthalmoscopy for the detection of clinically significant macular edema were 81% and 98%, respectively. Smartphone ophthalmoscopy and biomicroscopy could not be used to examine the fundus and grade DR in 9 eyes (3.75%) and 4 eyes (1.7%), respectively, because of cataract and/or small pupil diameter. Smartphone ophthalmoscopy showed considerable agreement with dilated retinal biomicroscopy for the grading of DR. The portability, affordability, and connectivity of a smartphone ophthalmoscope make smartphone ophthalmoscopy a promising technique for community screening programs.
Product Description
Supelco
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
R-(-)-Phenylephrine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
Tropicamide, solid
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Tropicamide for peak identification, European Pharmacopoeia (EP) Reference Standard
Tropicamide, European Pharmacopoeia (EP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard