Skip to Content
Merck
All Photos(1)

Key Documents

T2580000

Tropicamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-Ethyl-2-phenyl-N-(4-pyridylmethyl)hydracrylamide, Ro 1-7683

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O2
CAS Number:
Molecular Weight:
284.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tropicamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN(Cc1ccncc1)C(=O)C(CO)c2ccccc2

InChI

1S/C17H20N2O2/c1-2-19(12-14-8-10-18-11-9-14)17(21)16(13-20)15-6-4-3-5-7-15/h3-11,16,20H,2,12-13H2,1H3

InChI key

BGDKAVGWHJFAGW-UHFFFAOYSA-N

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tropicamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachael Cordina et al.
Clinical & experimental ophthalmology, 43(2), 115-123 (2014-08-01)
This study investigated the long-term ophthalmological consequences of cyanotic congenital heart disease (CHD). Cross-sectional study, tertiary referral setting. Thirteen adults with cyanotic CHD (40 ± 4 years). Age- and sex-matched healthy controls underwent aspects of the protocol. Cyanosed subjects had a full
Kalpana Rose et al.
Investigative ophthalmology & visual science, 55(7), 4266-4276 (2014-05-08)
The aim of this study was to investigate the impact of cigarette smoking in otherwise healthy young individuals on retinal blood flow (RBF) and vascular reactivity (RVR). An automated gas flow controller (RespirAct) was used to achieve normoxic hypercapnia in
Alja Crnej et al.
Investigative ophthalmology & visual science, 55(6), 3681-3685 (2014-05-17)
To establish a murine model for keratoprosthesis. A miniature keratoprosthesis (m-KPro) device was created consisting of a poly[methyl methacrylate] front part and a titanium back plate, designed after the Boston KPro, which is in widespread clinical use. BALB/c mice were
Anne E Powell et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(1), G16-G23 (2014-05-17)
Individuals with familial adenomatous polyposis (FAP) harbor a germline mutation in adenomatous polyposis coli (APC). The major clinical manifestation is development of multiple colonic tumors at a young age due to stochastic loss of the remaining APC allele. Extracolonic features
Xiaoyu Li et al.
Toxicology letters, 228(3), 200-206 (2014-05-14)
Acid-sensing ion channels (ASICs) are extracellular pH sensors activated by protons, which influence retinal activity and phototransduction. Among all ASICs, ASIC1a is abundantly expressed in the retina and involved in normal retinal activity. Chloroquine, which has been used in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service