Skip to Content
Merck
All Photos(1)

Key Documents

P-078

Supelco

R-(-)-Phenylephrine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14ClNO2
CAS Number:
Molecular Weight:
203.67
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CNC[C@H](O)c1cccc(O)c1

InChI

1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/t9-;/m0./s1

InChI key

OCYSGIYOVXAGKQ-FVGYRXGTSA-N

Gene Information

General description

Phenylephrine, marketed as Neo-Synephrine®, is the most common over-the-counter (OTC) decongestant in the US. Over the last few years, it has been marketed as a substitute for pseudoephedrine with tighter US government regulation of pseudoephedrine from the Combat Methamphetamine Epidemic Act of 2005.

Application

  • Stability-Indicating Assay for Ibuprofen and Phenylephrine hydrochloride: A high-performance liquid chromatography (HPLC) method was developed for the eco-friendly and stability-indicating assay of a binary mixture containing phenylephrine hydrochloride, highlighting its role in pharmaceutical quality control and formulation research (Kelani et al., 2023).
  • Visual Attention and Adrenergic Manipulation: The differential effects of dopaminergic and noradrenergic manipulations, including the use of phenylephrine, on attention were studied using the theory of visual attention in rat models, providing insights into the neuropharmacological impacts of adrenergic receptor agonists (Hervig et al., 2023).
  • Chemometric Quality Assessment: Comparative UV-spectrophotometric analysis using predictive models for the quality assessment of phenylephrine HCl and paracetamol, which is crucial for ensuring the chiral purity and efficacy of pharmaceutical formulations (Farid et al., 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Neo-Synephrine is a registered trademark of Bayer Healthcare LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie W Watts et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(6), 1320-1328 (2013-04-06)
Obesity and hypertension are comorbid in epidemic proportion, yet their biological connection is largely a mystery. The peptide chemerin is a candidate for connecting fat deposits around the blood vessel (perivascular adipose tissue) to arterial contraction. We presently tested the
Jin O-Uchi et al.
Biochemical and biophysical research communications, 433(2), 188-193 (2013-03-05)
Adrenoceptor stimulation is a key determinant of cardiac excitation-contraction coupling mainly through the activation of serine/threonine kinases. However, little is known about the role of protein tyrosine kinases (PTKs) activated by adrenergic signaling on cardiac excitation-contraction coupling. A cytoplasmic tyrosine
Abiodun Adefurin et al.
Hypertension (Dallas, Tex. : 1979), 61(4), 915-920 (2013-02-13)
Blacks have increased hemodynamic responses to both physiological and pharmacological adrenergic stimulation compared with whites, and this may contribute to the greater prevalence of hypertension in this ethnic group. A small study suggested enhanced α1-adrenoreceptor-mediated arterial vasoconstriction in the forearm
S Kauffmann et al.
Annales francaises d'anesthesie et de reanimation, 32(2), 112-114 (2013-02-05)
Adverse systemic reactions such as cardiovascular effects may occur following topical ocular application of phenylephrine. We report the case of a 49-year-old woman who, following topical application of phenylephrine eyedrops, experienced hypertension with electrocardiographic modifications and a rise in cardiac
Kyungjin Lee et al.
BMC complementary and alternative medicine, 13, 31-31 (2013-02-16)
Prunus yedoensis Matsum. is used as traditional medicine-'Yaeng-Pi' or 'Hua-Pi'-in Japan and Korea. However, no studies have examined the pharmacological activities of the P. yedoensis bark. Only the antioxidant and antiviral activities of P. yedoensis fruit and the anti-hyperglycaemic effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service