Merck
  • Short-chain fatty acids enhance adipocyte differentiation in the stromal vascular fraction of porcine adipose tissue.
Short-chain fatty acids enhance adipocyte differentiation in the stromal vascular fraction of porcine adipose tissue.
The Journal of nutrition (2014-10-17)
Genlai Li, Wen Yao, Honglin Jiang
ABSTRACT
Short-chain fatty acids (SCFAs), including acetate, propionate, and butyrate, are the main products of microbial fermentation in the gut and might mediate some of the effects of gut microbiota and nutrition on development, metabolism, and pathogenesis of obesity and other diseases. The objective of this study was to determine the effects of SCFAs on adipocyte differentiation and the underlying mechanism. The stromal vascular fraction (SVF) of the porcine subcutaneous fat was used as the preadipocyte model. Adipocyte differentiation was assessed by Oil Red O staining and gene expression analysis of adipocyte markers. Chromatin immunoprecipitation was used to assess the histone acetylation amounts at the peroxisome proliferator-activated receptor γ (PPARG) and CCAAT/enhancer binding protein α (CEBPA) promoters. Compared with control, propionate and butyrate enhanced the formation of adipocytes by 10-20% and mRNA expression of adipocyte markers by 20-200% in porcine SVF undergoing adipocyte differentiation. Compared with control, short-term treatment of propionate and butyrate enhanced PPARG and CEBPA mRNA expression in porcine SVF by 50-100%. Neither free fatty acid receptor (FFAR) 2 nor FFAR3 mRNA was detectable in porcine SVF before or during differentiation. Neither a cAMP analogue nor an activator of AMP-activated protein kinase (AMPK) affected propionate- or butyrate-enhanced expression of PPARG or CEBPA mRNA. Trichostatin A, a specific inhibitor of histone deacetylases (HDACs), enhanced the formation of adipocytes in porcine SVF by nearly 100% and the expression of PPARG and CEBPA mRNAs by 150% and 50%, respectively. Butyrate increased whereas propionate had no significant effect on histone H3 acetylation at the CEBPA promoter in porcine SVF. Propionate and butyrate enhance adipocyte differentiation in porcine SVF. These effects are unlikely mediated through FFAR2, FFAR3, cAMP, or AMPK. The effect of butyrate may be partially mediated by its HDAC inhibitory activity, whereas that of propionate is independent of its HDAC inhibitory activity.
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)
Sigma-Aldrich
Sodium propionate, FCC
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium propionate, ≥99.0%
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Supelco
Sodium propionate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications